Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75002964-75003237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4842345 c.132G>C p.Q44H Chr.14: 75003122-75003122 on Build GRCh38
6314202 c.76G>T p.G26C Chr.14: 75003066-75003066 on Build GRCh38
4519845 c.102G>C p.M34I Chr.14: 75003092-75003092 on Build GRCh38
3498057 c.45C>T p.I15I Chr.14: 75003035-75003035 on Build GRCh38
1300830 c.93C>T p.S31S Chr.14: 75003083-75003083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369180674 Chr.14: 75003067-75003067 on Build GRCh38
rs768335115 Chr.14: 75003145-75003145 on Build GRCh38
rs767933078 Chr.14: 75002993-75002993 on Build GRCh38
rs369798006 Chr.14: 75003171-75003171 on Build GRCh38
rs755897515 Chr.14: 75003107-75003107 on Build GRCh38
rs751443080 Chr.14: 75003020-75003020 on Build GRCh38
rs760542683 Chr.14: 75003154-75003154 on Build GRCh38
rs976260927 Chr.14: 75003148-75003148 on Build GRCh38
rs139325888 Chr.14: 75003109-75003109 on Build GRCh38
rs779230535 Chr.14: 75003188-75003188 on Build GRCh38
rs749552337 Chr.14: 75003041-75003041 on Build GRCh38
rs774957796 Chr.14: 75002989-75002989 on Build GRCh38
rs143091762 Chr.14: 75003054-75003054 on Build GRCh38
rs546027923 Chr.14: 75003207-75003207 on Build GRCh38
rs955152478 Chr.14: 75003169-75003169 on Build GRCh38
rs748288181 Chr.14: 75003126-75003126 on Build GRCh38
rs750307992 Chr.14: 75003153-75003153 on Build GRCh38
rs773695426 Chr.14: 75002995-75002995 on Build GRCh38
rs774585521 Chr.14: 75003053-75003053 on Build GRCh38
rs146228503 Chr.14: 75003086-75003086 on Build GRCh38
rs755230776 Chr.14: 75003187-75003187 on Build GRCh38
rs747542265 Chr.14: 75003202-75003202 on Build GRCh38
rs777041434 Chr.14: 75003141-75003141 on Build GRCh38
rs1038490990 Chr.14: 75003044-75003044 on Build GRCh38
rs773429046 Chr.14: 75003058-75003058 on Build GRCh38
rs761260701 Chr.14: 75003152-75003152 on Build GRCh38
rs771272417 Chr.14: 75003138-75003138 on Build GRCh38
rs985649534 Chr.14: 75003179-75003179 on Build GRCh38
rs768788808 Chr.14: 75003042-75003042 on Build GRCh38
rs377356154 Chr.14: 75003161-75003161 on Build GRCh38
rs772067107 Chr.14: 75003128-75003128 on Build GRCh38
rs759080911 Chr.14: 75002997-75002997 on Build GRCh38
rs754054549 Chr.14: 75003110-75003110 on Build GRCh38
rs776788187 Chr.14: 75003036-75003036 on Build GRCh38
rs941048370 Chr.14: 75003200-75003200 on Build GRCh38
rs747480025 Chr.14: 75003134-75003134 on Build GRCh38
rs757868212 Chr.14: 75003006-75003006 on Build GRCh38
rs780269972 Chr.14: 75003040-75003040 on Build GRCh38
rs767536443 Chr.14: 75003077-75003077 on Build GRCh38
rs780841869 Chr.14: 75003028-75003028 on Build GRCh38
rs553824476 Chr.14: 75003142-75003142 on Build GRCh38
rs957633719 Chr.14: 75003084-75003084 on Build GRCh38
rs529670879 Chr.14: 75003079-75003079 on Build GRCh38
rs780480791 Chr.14: 75003133-75003135 on Build GRCh38
rs762611298 Chr.14: 75003071-75003071 on Build GRCh38
rs753750841 Chr.14: 75003176-75003176 on Build GRCh38
rs761706101 Chr.14: 75003075-75003075 on Build GRCh38
rs199671360 Chr.14: 75002991-75002991 on Build GRCh38
rs141355163 Chr.14: 75003066-75003066 on Build GRCh38
rs144753546 Chr.14: 75003074-75003074 on Build GRCh38
rs752315506 Chr.14: 75003005-75003005 on Build GRCh38
rs764989690 Chr.14: 75003002-75003002 on Build GRCh38
rs779106104 Chr.14: 75003122-75003122 on Build GRCh38
rs372838502 Chr.14: 75003117-75003117 on Build GRCh38
rs772748876 Chr.14: 75003057-75003057 on Build GRCh38
rs757245015 Chr.14: 75003022-75003022 on Build GRCh38
rs747554501 Chr.14: 75003190-75003191 on Build GRCh38
rs763584691 Chr.14: 75003073-75003073 on Build GRCh38
rs766181401 Chr.14: 75003163-75003163 on Build GRCh38
rs148506549 Chr.14: 75003199-75003199 on Build GRCh38
rs778303902 Chr.14: 75003130-75003130 on Build GRCh38
rs745712923 Chr.14: 75003038-75003038 on Build GRCh38
rs773826562 Chr.14: 75003149-75003149 on Build GRCh38
rs756263862 Chr.14: 75003039-75003039 on Build GRCh38
rs1031494024 Chr.14: 75003156-75003156 on Build GRCh38
rs763602964 Chr.14: 75003007-75003007 on Build GRCh38
rs752999765 Chr.14: 75003194-75003194 on Build GRCh38
rs768878879 Chr.14: 75002986-75002986 on Build GRCh38
rs907307477 Chr.14: 75003013-75003013 on Build GRCh38

View Full Product Details