Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154988311-154988546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLAD1
Entrez Gene ID
80308
Gene Name
flavin adenine dinucleotide synthetase 1
Gene Aliases
FAD1, FADS, LSMFLAD, PP591
UniGene
Hs.118666
Gene Chromosome Location
Chr.1: 154983294-154993111 on Build GRCh38
Transcripts
NM_001184891.1 NM_001184892.1 NM_025207.4 NM_201398.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6664383 c.487C>T p.R163W Chr.1: 154988516-154988516 on Build GRCh38
4022898 c.751G>A p.V251I Chr.1: 154988483-154988483 on Build GRCh38
6664388 c.430T>C p.F144L Chr.1: 154988459-154988459 on Build GRCh38
269179 c.618A>G p.E206E Chr.1: 154988350-154988350 on Build GRCh38
7195729 c.344G>A p.G115E Chr.1: 154988373-154988373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148134209 Chr.1: 154988379-154988379 on Build GRCh38
rs879119618 Chr.1: 154988417-154988417 on Build GRCh38
rs148868251 Chr.1: 154988508-154988508 on Build GRCh38
rs751012776 Chr.1: 154988430-154988430 on Build GRCh38
rs776497411 Chr.1: 154988523-154988523 on Build GRCh38
rs144986636 Chr.1: 154988415-154988415 on Build GRCh38
rs1009078855 Chr.1: 154988389-154988389 on Build GRCh38
rs755481310 Chr.1: 154988435-154988435 on Build GRCh38
rs61736264 Chr.1: 154988461-154988461 on Build GRCh38
rs754464306 Chr.1: 154988487-154988487 on Build GRCh38
rs1041500319 Chr.1: 154988392-154988392 on Build GRCh38
rs566351390 Chr.1: 154988520-154988520 on Build GRCh38
rs370985969 Chr.1: 154988517-154988517 on Build GRCh38
rs769942679 Chr.1: 154988369-154988369 on Build GRCh38
rs776874328 Chr.1: 154988341-154988341 on Build GRCh38
rs199979286 Chr.1: 154988477-154988477 on Build GRCh38
rs138843652 Chr.1: 154988377-154988377 on Build GRCh38
rs757970182 Chr.1: 154988426-154988426 on Build GRCh38
rs572645053 Chr.1: 154988336-154988336 on Build GRCh38
rs752137994 Chr.1: 154988402-154988402 on Build GRCh38
rs146501091 Chr.1: 154988519-154988519 on Build GRCh38
rs764845268 Chr.1: 154988416-154988416 on Build GRCh38
rs750133322 Chr.1: 154988478-154988478 on Build GRCh38
rs145054820 Chr.1: 154988372-154988372 on Build GRCh38
rs772159163 Chr.1: 154988522-154988522 on Build GRCh38
rs761796690 Chr.1: 154988525-154988525 on Build GRCh38
rs989269955 Chr.1: 154988507-154988507 on Build GRCh38
rs150683998 Chr.1: 154988402-154988402 on Build GRCh38
rs766539386 Chr.1: 154988378-154988378 on Build GRCh38
rs748480567 Chr.1: 154988359-154988359 on Build GRCh38
rs749521512 Chr.1: 154988450-154988450 on Build GRCh38
rs764022484 Chr.1: 154988408-154988408 on Build GRCh38
rs777900563 Chr.1: 154988449-154988449 on Build GRCh38
rs771215649 Chr.1: 154988460-154988460 on Build GRCh38
rs770176496 Chr.1: 154988444-154988444 on Build GRCh38
rs753219806 Chr.1: 154988500-154988500 on Build GRCh38
rs779525564 Chr.1: 154988428-154988428 on Build GRCh38
rs759646371 Chr.1: 154988463-154988463 on Build GRCh38
rs374803472 Chr.1: 154988414-154988414 on Build GRCh38
rs145584366 Chr.1: 154988476-154988476 on Build GRCh38
rs765950156 Chr.1: 154988482-154988482 on Build GRCh38
rs1032884815 Chr.1: 154988446-154988446 on Build GRCh38
rs781696572 Chr.1: 154988441-154988441 on Build GRCh38
rs879171776 Chr.1: 154988338-154988338 on Build GRCh38
rs778273919 Chr.1: 154988515-154988515 on Build GRCh38
rs776606717 Chr.1: 154988474-154988474 on Build GRCh38
rs781601059 Chr.1: 154988499-154988499 on Build GRCh38
rs376550934 Chr.1: 154988516-154988516 on Build GRCh38
rs775640575 Chr.1: 154988380-154988380 on Build GRCh38
rs192926060 Chr.1: 154988483-154988483 on Build GRCh38
rs750044923 Chr.1: 154988421-154988421 on Build GRCh38
rs188177526 Chr.1: 154988473-154988473 on Build GRCh38

View Full Product Details