Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 51829434-51829705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCF3
Entrez Gene ID
401067
Gene Name
IQ motif containing F3
Gene Aliases
-
UniGene
Hs.729443
Gene Chromosome Location
Chr.3: 51826883-51830858 on Build GRCh38
Transcripts
NM_001085479.2 NM_001207023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3781845 c.58C>T p.R20W Chr.3: 51829704-51829704 on Build GRCh38
3595801 c.46G>A p.E16K Chr.3: 51829692-51829692 on Build GRCh38
2151667 c.59G>A p.R20Q Chr.3: 51829705-51829705 on Build GRCh38
6898807 c.22G>A p.G8S Chr.3: 51829668-51829668 on Build GRCh38
5821227 c.39T>A p.D13E Chr.3: 51829685-51829685 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010392159 Chr.3: 51829457-51829457 on Build GRCh38
rs781531071 Chr.3: 51829632-51829632 on Build GRCh38
rs371679655 Chr.3: 51829676-51829676 on Build GRCh38
rs370435748 Chr.3: 51829657-51829657 on Build GRCh38
rs760297595 Chr.3: 51829487-51829487 on Build GRCh38
rs907939568 Chr.3: 51829574-51829574 on Build GRCh38
rs373444918 Chr.3: 51829610-51829610 on Build GRCh38
rs765276230 Chr.3: 51829525-51829525 on Build GRCh38
rs766250105 Chr.3: 51829520-51829520 on Build GRCh38
rs377687328 Chr.3: 51829617-51829617 on Build GRCh38
rs777805886 Chr.3: 51829615-51829615 on Build GRCh38
rs757460164 Chr.3: 51829627-51829627 on Build GRCh38
rs183904433 Chr.3: 51829579-51829579 on Build GRCh38
rs767038067 Chr.3: 51829467-51829467 on Build GRCh38
rs758985211 Chr.3: 51829623-51829623 on Build GRCh38
rs754956850 Chr.3: 51829524-51829524 on Build GRCh38
rs1019650101 Chr.3: 51829476-51829476 on Build GRCh38
rs535839358 Chr.3: 51829593-51829593 on Build GRCh38
rs903706677 Chr.3: 51829502-51829502 on Build GRCh38
rs921947396 Chr.3: 51829528-51829528 on Build GRCh38
rs373170182 Chr.3: 51829485-51829485 on Build GRCh38
rs940552166 Chr.3: 51829575-51829575 on Build GRCh38
rs371101520 Chr.3: 51829643-51829643 on Build GRCh38
rs772940733 Chr.3: 51829486-51829486 on Build GRCh38
rs770769725 Chr.3: 51829664-51829664 on Build GRCh38
rs984864493 Chr.3: 51829573-51829573 on Build GRCh38
rs776508280 Chr.3: 51829671-51829671 on Build GRCh38
rs146326208 Chr.3: 51829674-51829674 on Build GRCh38
rs114638469 Chr.3: 51829649-51829649 on Build GRCh38
rs996400534 Chr.3: 51829684-51829684 on Build GRCh38
rs376109040 Chr.3: 51829529-51829529 on Build GRCh38
rs776009035 Chr.3: 51829645-51829645 on Build GRCh38
rs753754935 Chr.3: 51829522-51829522 on Build GRCh38
rs201435910 Chr.3: 51829531-51829531 on Build GRCh38
rs750948734 Chr.3: 51829536-51829536 on Build GRCh38
rs374619488 Chr.3: 51829659-51829659 on Build GRCh38
rs141452753 Chr.3: 51829656-51829656 on Build GRCh38

View Full Product Details