The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278511.1 | NP_001265440.1 | 21 - 22 | 2885 | 69 |
NM_001278512.1 | NP_001265441.1 | 23 - 24 | 3038 | 69 | |
NM_004644.4 | NP_004635.2 | 22 - 23 | 2981 | 69 | |
XM_017022640.1 | XP_016878129.1 | 21 - 22 | 3113 | 69 | |
GenBank mRNA | AF022152.1 | - | 22 - 23 | 2810 | 69 |
BC093739.1 | - | 22 - 23 | 2852 | 69 | |
BC143342.1 | - | 21 - 22 | 2756 | 69 | |
BC143346.1 | - | 23 - 24 | 2909 | 69 | |
CR749860.1 | - | 21 - 22 | 2980 | 69 | |
U37673.1 | - | 22 - 23 | 2837 | 69 |