Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFXL1
Entrez Gene ID
152518
Gene Name
nuclear transcription factor, X-box binding like 1
Gene Aliases
CDZFP, HOZFP, OZFP, URCC5
Chromosome Location
Chr.4: 47847233 - 47914667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278623.1 NP_001265552.1 9 - 10 1257 84
NM_001278624.1 NP_001265553.1 9 - 10 1395 84
NM_152995.5 NP_694540.3 9 - 10 1297 84
NR_103795.1 9 - 10 1257 84
GenBank mRNA AB085695.1 - 9 - 10 1253 84
AB181916.1 - 9 - 10 1377 84
AK131285.1 - 7 - 8 843 84
AK302440.1 - 9 - 10 1255 84
BC051193.1 - 9 - 10 1228 84

View Full Product Details