Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 14848681-14848952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA14
Entrez Gene ID
51182
Gene Name
heat shock protein family A (Hsp70) member 14
Gene Aliases
HSP70-4, HSP70L1
UniGene
Hs.534169
Gene Chromosome Location
Chr.10: 14838160-14871741 on Build GRCh38
Transcripts
NM_016299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
916599 c.329T>G p.F110C Chr.10: 14848848-14848848 on Build GRCh38
5516516 c.295C>T p.R99* Chr.10: 14848814-14848814 on Build GRCh38
7301459 c.313G>A p.G105R Chr.10: 14848832-14848832 on Build GRCh38
3382778 c.300T>C p.Y100Y Chr.10: 14848819-14848819 on Build GRCh38
1347010 c.278delA p.N95fs*8 Chr.10: 14848797-14848797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372812746 Chr.10: 14848835-14848835 on Build GRCh38
rs749748267 Chr.10: 14848768-14848768 on Build GRCh38
rs991975247 Chr.10: 14848736-14848737 on Build GRCh38
rs756876136 Chr.10: 14848840-14848840 on Build GRCh38
rs766422205 Chr.10: 14848815-14848815 on Build GRCh38
rs1044983161 Chr.10: 14848824-14848824 on Build GRCh38
rs45567440 Chr.10: 14848830-14848830 on Build GRCh38
rs780962885 Chr.10: 14848849-14848849 on Build GRCh38
rs745337056 Chr.10: 14848855-14848855 on Build GRCh38
rs762136566 Chr.10: 14848829-14848829 on Build GRCh38
rs753850992 Chr.10: 14848816-14848816 on Build GRCh38
rs201058008 Chr.10: 14848823-14848823 on Build GRCh38
rs547021710 Chr.10: 14848914-14848914 on Build GRCh38
rs761157024 Chr.10: 14848913-14848913 on Build GRCh38
rs760011797 Chr.10: 14848796-14848797 on Build GRCh38
rs913710404 Chr.10: 14848741-14848741 on Build GRCh38
rs377076504 Chr.10: 14848789-14848789 on Build GRCh38
rs773524564 Chr.10: 14848794-14848794 on Build GRCh38
rs776010439 Chr.10: 14848748-14848748 on Build GRCh38
rs879056361 Chr.10: 14848729-14848729 on Build GRCh38
rs910911909 Chr.10: 14848713-14848713 on Build GRCh38
rs773722919 Chr.10: 14848908-14848908 on Build GRCh38
rs765131686 Chr.10: 14848822-14848822 on Build GRCh38
rs111362845 Chr.10: 14848871-14848871 on Build GRCh38
rs749426152 Chr.10: 14848753-14848753 on Build GRCh38
rs777231181 Chr.10: 14848778-14848782 on Build GRCh38
rs774949480 Chr.10: 14848873-14848873 on Build GRCh38
rs375853738 Chr.10: 14848872-14848872 on Build GRCh38
rs34106206 Chr.10: 14848882-14848882 on Build GRCh38
rs570960603 Chr.10: 14848821-14848821 on Build GRCh38
rs548363926 Chr.10: 14848717-14848717 on Build GRCh38
rs935056133 Chr.10: 14848785-14848785 on Build GRCh38
rs190498380 Chr.10: 14848772-14848772 on Build GRCh38
rs550628295 Chr.10: 14848798-14848798 on Build GRCh38
rs768016540 Chr.10: 14848790-14848790 on Build GRCh38
rs976357952 Chr.10: 14848722-14848722 on Build GRCh38
rs758140227 Chr.10: 14848827-14848827 on Build GRCh38
rs561899709 Chr.10: 14848720-14848720 on Build GRCh38
rs750700184 Chr.10: 14848850-14848850 on Build GRCh38
rs748692573 Chr.10: 14848884-14848884 on Build GRCh38

View Full Product Details