Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35548629-35548890 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC3
Entrez Gene ID
51614
Gene Name
ERGIC and golgi 3
Gene Aliases
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
UniGene
Hs.472558
Gene Chromosome Location
Chr.20: 35542029-35557634 on Build GRCh38
Transcripts
NM_015966.2 NM_198398.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7090210 c.655G>T p.G219W Chr.20: 35548835-35548835 on Build GRCh38
6656089 c.634G>A p.G212R Chr.20: 35548814-35548814 on Build GRCh38
4291585 c.634G>A p.G212R Chr.20: 35548814-35548814 on Build GRCh38
6258788 c.657G>T p.G219G Chr.20: 35548837-35548837 on Build GRCh38
1026255 c.604T>C p.Y202H Chr.20: 35548651-35548651 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752814923 Chr.20: 35548655-35548655 on Build GRCh38
rs747829579 Chr.20: 35548665-35548665 on Build GRCh38
rs760886960 Chr.20: 35548698-35548698 on Build GRCh38
rs762724761 Chr.20: 35548677-35548677 on Build GRCh38
rs759620258 Chr.20: 35548690-35548690 on Build GRCh38
rs777629170 Chr.20: 35548777-35548777 on Build GRCh38
rs921160779 Chr.20: 35548741-35548741 on Build GRCh38
rs757609562 Chr.20: 35548709-35548709 on Build GRCh38
rs764276399 Chr.20: 35548846-35548846 on Build GRCh38
rs1054657718 Chr.20: 35548744-35548744 on Build GRCh38
rs764363895 Chr.20: 35548701-35548701 on Build GRCh38
rs976680854 Chr.20: 35548727-35548727 on Build GRCh38
rs780955750 Chr.20: 35548659-35548659 on Build GRCh38
rs760358675 Chr.20: 35548861-35548861 on Build GRCh38
rs770660482 Chr.20: 35548790-35548790 on Build GRCh38
rs202155461 Chr.20: 35548715-35548715 on Build GRCh38
rs770890066 Chr.20: 35548685-35548685 on Build GRCh38
rs182209758 Chr.20: 35548671-35548671 on Build GRCh38
rs934492507 Chr.20: 35548735-35548735 on Build GRCh38
rs749088943 Chr.20: 35548789-35548789 on Build GRCh38
rs893060760 Chr.20: 35548805-35548805 on Build GRCh38
rs779446825 Chr.20: 35548713-35548713 on Build GRCh38
rs754185898 Chr.20: 35548705-35548705 on Build GRCh38
rs867087855 Chr.20: 35548731-35548731 on Build GRCh38
rs200704504 Chr.20: 35548864-35548864 on Build GRCh38
rs774786739 Chr.20: 35548786-35548787 on Build GRCh38
rs932308355 Chr.20: 35548814-35548814 on Build GRCh38
rs769569948 Chr.20: 35548776-35548776 on Build GRCh38
rs752858435 Chr.20: 35548697-35548697 on Build GRCh38
rs760800923 Chr.20: 35548837-35548837 on Build GRCh38
rs747739875 Chr.20: 35548775-35548775 on Build GRCh38
rs769655761 Chr.20: 35548669-35548669 on Build GRCh38
rs74834954 Chr.20: 35548762-35548762 on Build GRCh38
rs775543043 Chr.20: 35548833-35548833 on Build GRCh38
rs144274449 Chr.20: 35548826-35548826 on Build GRCh38
rs754049984 Chr.20: 35548858-35548858 on Build GRCh38
rs532717308 Chr.20: 35548771-35548771 on Build GRCh38
rs767605074 Chr.20: 35548696-35548696 on Build GRCh38
rs774292745 Chr.20: 35548796-35548796 on Build GRCh38
rs754462476 Chr.20: 35548722-35548722 on Build GRCh38
rs146493836 Chr.20: 35548831-35548831 on Build GRCh38

View Full Product Details