Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153455461-153455686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS7
Entrez Gene ID
55559
Gene Name
HAUS augmin like complex subunit 7
Gene Aliases
UCHL5IP, UIP1
UniGene
Hs.170835
Gene Chromosome Location
Chr.X: 153447665-153495525 on Build GRCh38
Transcripts
NM_017518.7 NR_073156.1

SNP Detail

SNP ID Chromosome Location
rs782096283 Chr.X: 153455519-153455519 on Build GRCh38
rs375613209 Chr.X: 153455628-153455628 on Build GRCh38
rs1015781747 Chr.X: 153455533-153455533 on Build GRCh38
rs782168710 Chr.X: 153455621-153455621 on Build GRCh38
rs782682700 Chr.X: 153455616-153455616 on Build GRCh38
rs782245548 Chr.X: 153455500-153455500 on Build GRCh38
rs781919065 Chr.X: 153455662-153455662 on Build GRCh38
rs781996330 Chr.X: 153455532-153455532 on Build GRCh38
rs782597035 Chr.X: 153455629-153455629 on Build GRCh38
rs1048670086 Chr.X: 153455495-153455495 on Build GRCh38
rs781885664 Chr.X: 153455559-153455559 on Build GRCh38
rs781915818 Chr.X: 153455517-153455517 on Build GRCh38
rs143438621 Chr.X: 153455593-153455593 on Build GRCh38
rs782258428 Chr.X: 153455637-153455637 on Build GRCh38
rs901316353 Chr.X: 153455568-153455568 on Build GRCh38
rs782298923 Chr.X: 153455511-153455511 on Build GRCh38
rs782109433 Chr.X: 153455536-153455536 on Build GRCh38
rs782257325 Chr.X: 153455490-153455490 on Build GRCh38
rs148815139 Chr.X: 153455590-153455590 on Build GRCh38
rs781841239 Chr.X: 153455583-153455583 on Build GRCh38
rs782621030 Chr.X: 153455500-153455500 on Build GRCh38
rs371952440 Chr.X: 153455641-153455641 on Build GRCh38
rs368229906 Chr.X: 153455638-153455638 on Build GRCh38
rs373704500 Chr.X: 153455512-153455512 on Build GRCh38
rs782443125 Chr.X: 153455497-153455497 on Build GRCh38
rs781990821 Chr.X: 153455510-153455510 on Build GRCh38
rs782818750 Chr.X: 153455551-153455551 on Build GRCh38
rs6571304 Chr.X: 153455521-153455521 on Build GRCh38
rs782321995 Chr.X: 153455643-153455643 on Build GRCh38
rs782221393 Chr.X: 153455502-153455502 on Build GRCh38
rs782701322 Chr.X: 153455580-153455580 on Build GRCh38
rs782399793 Chr.X: 153455508-153455508 on Build GRCh38
rs201262645 Chr.X: 153455570-153455570 on Build GRCh38
rs782033231 Chr.X: 153455640-153455640 on Build GRCh38

View Full Product Details