The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_197941.3 | NP_922932.2 | 25 - 25 | 7154 | 173 |
NR_135689.1 | 26 - 26 | 7305 | 173 | ||
XM_011543113.2 | XP_011541415.1 | 25 - 25 | 7169 | 173 | |
XM_011543114.2 | XP_011541416.1 | 25 - 25 | 6748 | 173 | |
XM_011543115.2 | XP_011541417.1 | 24 - 24 | 7057 | 173 | |
XM_011543116.2 | XP_011541418.1 | 23 - 23 | 6916 | 173 | |
XM_011543117.1 | XP_011541419.1 | 23 - 23 | 6507 | 173 | |
XM_011543121.2 | XP_011541423.2 | 21 - 21 | 5535 | 173 | |
XM_011543123.2 | XP_011541425.1 | 19 - 19 | 5273 | 173 | |
XM_011543124.2 | XP_011541426.1 | 16 - 16 | 5057 | 173 | |
XM_011543125.1 | XP_011541427.1 | 15 - 15 | 4912 | 173 | |
GenBank mRNA | AB209685.1 | - | 26 - 26 | 6802 | 173 |
AY692423.1 | - | 6 - 6 | 3757 | 173 | |
AY692424.1 | - | 25 - 25 | 7112 | 173 | |
BC010563.1 | - | 1 - 1 | 1305 | 173 | |
BC071654.1 | - | 1 - 1 | 1558 | 173 | |
CR749356.1 | - | 7 - 7 | 3889 | 173 |