Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 202035268-202035517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD7
Entrez Gene ID
8324
Gene Name
frizzled class receptor 7
Gene Aliases
FzE3
UniGene
Hs.173859
Gene Chromosome Location
Chr.2: 202034587-202038437 on Build GRCh38
Transcripts
NM_003507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2907809 c.803C>T p.T268M Chr.2: 202035450-202035450 on Build GRCh38
4090285 c.751C>T p.R251C Chr.2: 202035398-202035398 on Build GRCh38
5058566 c.761G>A p.R254H Chr.2: 202035408-202035408 on Build GRCh38
4090286 c.786G>A p.V262V Chr.2: 202035433-202035433 on Build GRCh38
4090283 c.668G>A p.R223H Chr.2: 202035315-202035315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778438593 Chr.2: 202035383-202035383 on Build GRCh38
rs935549711 Chr.2: 202035384-202035386 on Build GRCh38
rs759263156 Chr.2: 202035416-202035416 on Build GRCh38
rs766212417 Chr.2: 202035322-202035322 on Build GRCh38
rs376129245 Chr.2: 202035364-202035364 on Build GRCh38
rs140897513 Chr.2: 202035410-202035410 on Build GRCh38
rs267599158 Chr.2: 202035500-202035500 on Build GRCh38
rs761282146 Chr.2: 202035355-202035365 on Build GRCh38
rs1052639191 Chr.2: 202035385-202035385 on Build GRCh38
rs747655093 Chr.2: 202035386-202035386 on Build GRCh38
rs771504922 Chr.2: 202035388-202035388 on Build GRCh38
rs768155755 Chr.2: 202035448-202035448 on Build GRCh38
rs549284807 Chr.2: 202035430-202035430 on Build GRCh38
rs771979564 Chr.2: 202035298-202035298 on Build GRCh38
rs370505613 Chr.2: 202035372-202035372 on Build GRCh38
rs760416346 Chr.2: 202035313-202035313 on Build GRCh38
rs773165797 Chr.2: 202035299-202035299 on Build GRCh38
rs11546413 Chr.2: 202035491-202035491 on Build GRCh38
rs561680999 Chr.2: 202035315-202035315 on Build GRCh38
rs762470360 Chr.2: 202035445-202035445 on Build GRCh38
rs776801836 Chr.2: 202035341-202035341 on Build GRCh38
rs371541002 Chr.2: 202035459-202035459 on Build GRCh38
rs145569954 Chr.2: 202035346-202035346 on Build GRCh38
rs776466891 Chr.2: 202035415-202035415 on Build GRCh38
rs768050210 Chr.2: 202035286-202035286 on Build GRCh38
rs746990509 Chr.2: 202035403-202035403 on Build GRCh38
rs781734323 Chr.2: 202035370-202035370 on Build GRCh38
rs955853592 Chr.2: 202035495-202035495 on Build GRCh38
rs199969139 Chr.2: 202035466-202035466 on Build GRCh38
rs369157584 Chr.2: 202035449-202035449 on Build GRCh38
rs752131418 Chr.2: 202035359-202035359 on Build GRCh38
rs374592078 Chr.2: 202035395-202035395 on Build GRCh38
rs964029248 Chr.2: 202035406-202035406 on Build GRCh38
rs757874303 Chr.2: 202035477-202035477 on Build GRCh38
rs373396669 Chr.2: 202035345-202035345 on Build GRCh38
rs752110813 Chr.2: 202035436-202035436 on Build GRCh38
rs754605178 Chr.2: 202035382-202035382 on Build GRCh38
rs868385707 Chr.2: 202035408-202035408 on Build GRCh38
rs375789778 Chr.2: 202035484-202035484 on Build GRCh38
rs764652531 Chr.2: 202035355-202035355 on Build GRCh38
rs762942182 Chr.2: 202035354-202035354 on Build GRCh38
rs773973498 Chr.2: 202035291-202035291 on Build GRCh38
rs769990859 Chr.2: 202035422-202035422 on Build GRCh38
rs777668237 Chr.2: 202035400-202035400 on Build GRCh38
rs778579874 Chr.2: 202035461-202035461 on Build GRCh38
rs747629174 Chr.2: 202035293-202035293 on Build GRCh38

View Full Product Details