Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113844376-113844601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1C
Entrez Gene ID
54762
Gene Name
GRAM domain containing 1C
Gene Aliases
-
UniGene
Hs.24583
Gene Chromosome Location
Chr.3: 113828182-113947174 on Build GRCh38
Transcripts
NM_017577.4 XM_005247546.2 XM_005247547.1 XM_011512930.1 XM_017006646.1 XM_017006647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036565 c.61G>A p.D21N Chr.3: 113844536-113844536 on Build GRCh38
4642710 c.74A>G p.D25G Chr.3: 113844549-113844549 on Build GRCh38
4980282 c.108G>C p.E36D Chr.3: 113844583-113844583 on Build GRCh38
727152 c.89C>G p.P30R Chr.3: 113844564-113844564 on Build GRCh38
1308419 c.28G>A p.V10M Chr.3: 113844503-113844503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761448882 Chr.3: 113844508-113844508 on Build GRCh38
rs141093598 Chr.3: 113844534-113844534 on Build GRCh38
rs747144071 Chr.3: 113844476-113844476 on Build GRCh38
rs548225853 Chr.3: 113844550-113844550 on Build GRCh38
rs559302956 Chr.3: 113844467-113844467 on Build GRCh38
rs753696296 Chr.3: 113844536-113844536 on Build GRCh38
rs990045004 Chr.3: 113844409-113844409 on Build GRCh38
rs781684434 Chr.3: 113844557-113844557 on Build GRCh38
rs766090096 Chr.3: 113844535-113844535 on Build GRCh38
rs761056377 Chr.3: 113844421-113844421 on Build GRCh38
rs750349258 Chr.3: 113844513-113844513 on Build GRCh38
rs577453072 Chr.3: 113844475-113844475 on Build GRCh38
rs748380915 Chr.3: 113844455-113844455 on Build GRCh38
rs746367958 Chr.3: 113844564-113844564 on Build GRCh38
rs373447299 Chr.3: 113844420-113844420 on Build GRCh38
rs778013054 Chr.3: 113844547-113844547 on Build GRCh38
rs758402170 Chr.3: 113844457-113844457 on Build GRCh38
rs932196536 Chr.3: 113844450-113844450 on Build GRCh38
rs777038032 Chr.3: 113844486-113844486 on Build GRCh38
rs777821947 Chr.3: 113844464-113844464 on Build GRCh38
rs202089640 Chr.3: 113844569-113844569 on Build GRCh38
rs757309054 Chr.3: 113844551-113844551 on Build GRCh38
rs771442052 Chr.3: 113844479-113844479 on Build GRCh38
rs767446669 Chr.3: 113844511-113844511 on Build GRCh38
rs531924957 Chr.3: 113844491-113844491 on Build GRCh38
rs866572967 Chr.3: 113844505-113844505 on Build GRCh38
rs763898538 Chr.3: 113844501-113844501 on Build GRCh38
rs200881985 Chr.3: 113844489-113844489 on Build GRCh38
rs890597718 Chr.3: 113844485-113844485 on Build GRCh38
rs758603323 Chr.3: 113844545-113844545 on Build GRCh38
rs562411629 Chr.3: 113844503-113844503 on Build GRCh38
rs200526806 Chr.3: 113844499-113844499 on Build GRCh38
rs1051136709 Chr.3: 113844472-113844472 on Build GRCh38

View Full Product Details