Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 112418779-112419014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSDL2
Entrez Gene ID
84263
Gene Name
hydroxysteroid dehydrogenase like 2
Gene Aliases
C9orf99, SDR13C1
UniGene
Hs.59486
Gene Chromosome Location
Chr.9: 112379292-112472410 on Build GRCh38
Transcripts
NM_001195822.1 NM_032303.4 NR_036651.1 XM_011519091.2 XM_017015203.1 XM_017015204.1 XM_017015205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3996243 c.533A>G p.Y178C Chr.9: 112418893-112418893 on Build GRCh38
6397726 c.598G>T p.A200S Chr.9: 112418958-112418958 on Build GRCh38
1104143 c.553G>T p.E185* Chr.9: 112418913-112418913 on Build GRCh38
7093326 c.593A>C p.K198T Chr.9: 112418953-112418953 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770969333 Chr.9: 112418837-112418837 on Build GRCh38
rs892614209 Chr.9: 112418874-112418874 on Build GRCh38
rs746029771 Chr.9: 112418951-112418951 on Build GRCh38
rs200878102 Chr.9: 112418891-112418891 on Build GRCh38
rs772266593 Chr.9: 112418980-112418980 on Build GRCh38
rs753108158 Chr.9: 112418914-112418914 on Build GRCh38
rs964820790 Chr.9: 112418884-112418884 on Build GRCh38
rs956012968 Chr.9: 112418947-112418947 on Build GRCh38
rs750050506 Chr.9: 112418890-112418890 on Build GRCh38
rs373712932 Chr.9: 112418932-112418932 on Build GRCh38
rs1007556948 Chr.9: 112418956-112418956 on Build GRCh38
rs761363617 Chr.9: 112418882-112418882 on Build GRCh38
rs549611931 Chr.9: 112418879-112418879 on Build GRCh38
rs370811576 Chr.9: 112418898-112418898 on Build GRCh38
rs778867543 Chr.9: 112418950-112418950 on Build GRCh38
rs749360910 Chr.9: 112418827-112418827 on Build GRCh38
rs771106429 Chr.9: 112418834-112418836 on Build GRCh38
rs549386542 Chr.9: 112418888-112418888 on Build GRCh38
rs925985533 Chr.9: 112418946-112418946 on Build GRCh38
rs746111133 Chr.9: 112418847-112418847 on Build GRCh38
rs760516939 Chr.9: 112418863-112418863 on Build GRCh38
rs754328151 Chr.9: 112418937-112418937 on Build GRCh38
rs768572789 Chr.9: 112418871-112418871 on Build GRCh38
rs747219815 Chr.9: 112418828-112418832 on Build GRCh38
rs775275390 Chr.9: 112418851-112418851 on Build GRCh38
rs766194950 Chr.9: 112418894-112418894 on Build GRCh38
rs778324705 Chr.9: 112418936-112418936 on Build GRCh38
rs868805737 Chr.9: 112418941-112418941 on Build GRCh38
rs377111864 Chr.9: 112418945-112418945 on Build GRCh38
rs779149490 Chr.9: 112418841-112418841 on Build GRCh38
rs111704739 Chr.9: 112418850-112418850 on Build GRCh38

View Full Product Details