Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 100422958-100423229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMCN
Entrez Gene ID
51705
Gene Name
endomucin
Gene Aliases
EMCN2, MUC14
UniGene
Hs.152913
Gene Chromosome Location
Chr.4: 100395341-100518093 on Build GRCh38
Transcripts
NM_001159694.1 NM_016242.3 XM_011532024.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
334564 c.551C>A p.T184N Chr.4: 100423038-100423038 on Build GRCh38
5908393 c.527G>A p.G176E Chr.4: 100423062-100423062 on Build GRCh38
6098811 c.535G>A p.A179T Chr.4: 100423054-100423054 on Build GRCh38
5931475 c.567C>T p.S189S Chr.4: 100423022-100423022 on Build GRCh38
6206212 c.556C>T p.R186W Chr.4: 100423033-100423033 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189275756 Chr.4: 100422996-100422996 on Build GRCh38
rs771694055 Chr.4: 100423091-100423092 on Build GRCh38
rs775627583 Chr.4: 100423088-100423088 on Build GRCh38
rs562568224 Chr.4: 100423166-100423166 on Build GRCh38
rs751272792 Chr.4: 100423019-100423019 on Build GRCh38
rs2276935 Chr.4: 100423098-100423098 on Build GRCh38
rs776685234 Chr.4: 100422991-100422991 on Build GRCh38
rs762691920 Chr.4: 100423005-100423005 on Build GRCh38
rs772958309 Chr.4: 100423087-100423087 on Build GRCh38
rs775315481 Chr.4: 100423067-100423067 on Build GRCh38
rs116786214 Chr.4: 100423193-100423193 on Build GRCh38
rs767257795 Chr.4: 100423084-100423084 on Build GRCh38
rs756163730 Chr.4: 100423132-100423132 on Build GRCh38
rs765155216 Chr.4: 100423181-100423181 on Build GRCh38
rs988174767 Chr.4: 100423159-100423159 on Build GRCh38
rs775398832 Chr.4: 100423002-100423002 on Build GRCh38
rs189201905 Chr.4: 100423186-100423186 on Build GRCh38
rs753199148 Chr.4: 100423039-100423039 on Build GRCh38
rs888617372 Chr.4: 100423170-100423170 on Build GRCh38
rs771001270 Chr.4: 100422987-100422987 on Build GRCh38
rs141262429 Chr.4: 100423082-100423082 on Build GRCh38
rs375179838 Chr.4: 100423022-100423022 on Build GRCh38
rs749941433 Chr.4: 100423027-100423027 on Build GRCh38
rs779319831 Chr.4: 100423033-100423033 on Build GRCh38
rs369310114 Chr.4: 100423041-100423041 on Build GRCh38
rs181461181 Chr.4: 100423116-100423116 on Build GRCh38
rs375298461 Chr.4: 100423129-100423129 on Build GRCh38
rs775071677 Chr.4: 100423092-100423092 on Build GRCh38
rs755592173 Chr.4: 100423032-100423032 on Build GRCh38
rs768342617 Chr.4: 100423076-100423076 on Build GRCh38
rs767204719 Chr.4: 100423023-100423023 on Build GRCh38
rs778130632 Chr.4: 100423043-100423043 on Build GRCh38
rs753147720 Chr.4: 100423095-100423095 on Build GRCh38
rs757454828 Chr.4: 100423056-100423056 on Build GRCh38
rs138871242 Chr.4: 100423065-100423065 on Build GRCh38
rs759523556 Chr.4: 100422993-100422993 on Build GRCh38
rs774285293 Chr.4: 100423081-100423081 on Build GRCh38
rs764498761 Chr.4: 100423114-100423114 on Build GRCh38
rs376741806 Chr.4: 100422981-100422981 on Build GRCh38
rs1015858861 Chr.4: 100423187-100423187 on Build GRCh38
rs377524110 Chr.4: 100423105-100423105 on Build GRCh38
rs765854832 Chr.4: 100423091-100423091 on Build GRCh38
rs749194605 Chr.4: 100423074-100423074 on Build GRCh38
rs184846435 Chr.4: 100423128-100423128 on Build GRCh38
rs769880520 Chr.4: 100423066-100423066 on Build GRCh38
rs1006110454 Chr.4: 100423171-100423171 on Build GRCh38
rs757507825 Chr.4: 100423118-100423118 on Build GRCh38
rs373667023 Chr.4: 100423012-100423012 on Build GRCh38
rs758937068 Chr.4: 100423106-100423106 on Build GRCh38
rs979720917 Chr.4: 100423191-100423191 on Build GRCh38
rs561763267 Chr.4: 100423150-100423150 on Build GRCh38
rs747295104 Chr.4: 100423055-100423055 on Build GRCh38

View Full Product Details