Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138510476-138510727 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETF1
Entrez Gene ID
2107
Gene Name
eukaryotic translation termination factor 1
Gene Aliases
D5S1995, ERF, ERF1, RF1, SUP45L1, TB3-1
UniGene
Hs.483494
Gene Chromosome Location
Chr.5: 138506093-138543300 on Build GRCh38
Transcripts
NM_001256302.1 NM_001282185.1 NM_001291974.1 NM_001291975.1 NM_004730.3 XM_005271921.1 XM_017009221.1 XM_017009222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6335811 c.1083+1G>T p.? Chr.5: 138510564-138510564 on Build GRCh38
6656609 c.1070T>G p.F357C Chr.5: 138510578-138510578 on Build GRCh38
1061401 c.1051G>T p.E351* Chr.5: 138510597-138510597 on Build GRCh38
4818405 c.1051G>A p.E351K Chr.5: 138510597-138510597 on Build GRCh38
1433398 c.1031T>C p.L344P Chr.5: 138510617-138510617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770676071 Chr.5: 138510558-138510558 on Build GRCh38
rs745360280 Chr.5: 138510598-138510598 on Build GRCh38
rs762254263 Chr.5: 138510518-138510518 on Build GRCh38
rs911398576 Chr.5: 138510636-138510636 on Build GRCh38
rs1050409792 Chr.5: 138510628-138510628 on Build GRCh38
rs760923330 Chr.5: 138510654-138510654 on Build GRCh38
rs144420280 Chr.5: 138510533-138510533 on Build GRCh38
rs758215436 Chr.5: 138510546-138510546 on Build GRCh38
rs1006139533 Chr.5: 138510697-138510697 on Build GRCh38
rs764659540 Chr.5: 138510517-138510517 on Build GRCh38
rs375775652 Chr.5: 138510667-138510667 on Build GRCh38
rs944170614 Chr.5: 138510640-138510640 on Build GRCh38
rs747976555 Chr.5: 138510545-138510545 on Build GRCh38
rs756675629 Chr.5: 138510654-138510654 on Build GRCh38
rs756284629 Chr.5: 138510532-138510532 on Build GRCh38
rs769129789 Chr.5: 138510633-138510633 on Build GRCh38
rs750678167 Chr.5: 138510526-138510526 on Build GRCh38
rs754972400 Chr.5: 138510534-138510534 on Build GRCh38
rs948262327 Chr.5: 138510653-138510653 on Build GRCh38
rs186112104 Chr.5: 138510525-138510525 on Build GRCh38
rs765389836 Chr.5: 138510644-138510644 on Build GRCh38
rs562981902 Chr.5: 138510693-138510693 on Build GRCh38
rs748611176 Chr.5: 138510548-138510548 on Build GRCh38
rs368681169 Chr.5: 138510554-138510554 on Build GRCh38
rs963498500 Chr.5: 138510574-138510574 on Build GRCh38
rs1029927839 Chr.5: 138510507-138510507 on Build GRCh38
rs140291848 Chr.5: 138510702-138510702 on Build GRCh38
rs201296360 Chr.5: 138510652-138510652 on Build GRCh38
rs576212787 Chr.5: 138510670-138510670 on Build GRCh38
rs774929840 Chr.5: 138510639-138510639 on Build GRCh38
rs762189724 Chr.5: 138510642-138510642 on Build GRCh38
rs777285855 Chr.5: 138510502-138510502 on Build GRCh38
rs776028968 Chr.5: 138510703-138510703 on Build GRCh38
rs146595337 Chr.5: 138510689-138510689 on Build GRCh38

View Full Product Details