Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89632761-89633018 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF7
Entrez Gene ID
374654
Gene Name
kinesin family member 7
Gene Aliases
ACLS, AGBK, HLS2, JBTS12, UNQ340
UniGene
Hs.513134
Gene Chromosome Location
Chr.15: 89627970-89663086 on Build GRCh38
Transcripts
NM_198525.2 XM_011521531.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6695698 c.1315_1317delGAG p.E439delE Chr.15: 89632859-89632861 on Build GRCh38
255253 c.1295_1297delAGA p.K433delK Chr.15: 89632879-89632881 on Build GRCh38
4771489 c.1334G>T p.S445I Chr.15: 89632842-89632842 on Build GRCh38
6695709 c.2760G>T p.E920D Chr.15: 89632955-89632955 on Build GRCh38
6198413 c.2776C>T p.R926W Chr.15: 89632939-89632939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746577567 Chr.15: 89632957-89632957 on Build GRCh38
rs1043554942 Chr.15: 89632806-89632806 on Build GRCh38
rs764218371 Chr.15: 89632932-89632932 on Build GRCh38
rs758208567 Chr.15: 89632788-89632789 on Build GRCh38
rs747746877 Chr.15: 89632874-89632874 on Build GRCh38
rs567836651 Chr.15: 89632828-89632828 on Build GRCh38
rs556977418 Chr.15: 89632915-89632915 on Build GRCh38
rs556953796 Chr.15: 89632884-89632884 on Build GRCh38
rs781025699 Chr.15: 89632814-89632814 on Build GRCh38
rs369827484 Chr.15: 89632875-89632875 on Build GRCh38
rs371062109 Chr.15: 89632812-89632813 on Build GRCh38
rs3803530 Chr.15: 89632842-89632842 on Build GRCh38
rs569730394 Chr.15: 89632811-89632811 on Build GRCh38
rs147407072 Chr.15: 89632939-89632939 on Build GRCh38
rs753737326 Chr.15: 89632938-89632938 on Build GRCh38
rs867222588 Chr.15: 89632963-89632963 on Build GRCh38
rs879130956 Chr.15: 89632850-89632850 on Build GRCh38
rs757264957 Chr.15: 89632935-89632935 on Build GRCh38
rs771525697 Chr.15: 89632899-89632899 on Build GRCh38
rs781356957 Chr.15: 89632859-89632861 on Build GRCh38
rs761354914 Chr.15: 89632835-89632835 on Build GRCh38
rs746321431 Chr.15: 89632967-89632967 on Build GRCh38
rs138410620 Chr.15: 89632872-89632872 on Build GRCh38
rs779269518 Chr.15: 89632805-89632805 on Build GRCh38
rs746496879 Chr.15: 89632891-89632891 on Build GRCh38
rs766652877 Chr.15: 89632837-89632837 on Build GRCh38
rs769890385 Chr.15: 89632976-89632978 on Build GRCh38
rs749134116 Chr.15: 89632870-89632870 on Build GRCh38
rs372338933 Chr.15: 89632945-89632945 on Build GRCh38
rs372233228 Chr.15: 89632879-89632879 on Build GRCh38
rs758433714 Chr.15: 89632852-89632852 on Build GRCh38
rs947433457 Chr.15: 89632979-89632979 on Build GRCh38
rs1006371075 Chr.15: 89632907-89632907 on Build GRCh38
rs776064132 Chr.15: 89632977-89632977 on Build GRCh38
rs753784742 Chr.15: 89632992-89632992 on Build GRCh38
rs760976679 Chr.15: 89632782-89632782 on Build GRCh38
rs774408412 Chr.15: 89632982-89632982 on Build GRCh38
rs537333815 Chr.15: 89632812-89632812 on Build GRCh38
rs779835765 Chr.15: 89632823-89632823 on Build GRCh38
rs766347192 Chr.15: 89632936-89632936 on Build GRCh38
rs375379282 Chr.15: 89632869-89632869 on Build GRCh38
rs770287798 Chr.15: 89632976-89632976 on Build GRCh38
rs748140280 Chr.15: 89632877-89632879 on Build GRCh38
rs745819291 Chr.15: 89632789-89632803 on Build GRCh38
rs200502068 Chr.15: 89632794-89632794 on Build GRCh38
rs769493560 Chr.15: 89632817-89632817 on Build GRCh38
rs750332974 Chr.15: 89632856-89632856 on Build GRCh38
rs761986869 Chr.15: 89632983-89632983 on Build GRCh38
rs775869714 Chr.15: 89632928-89632928 on Build GRCh38
rs778564914 Chr.15: 89632873-89632873 on Build GRCh38
rs964966183 Chr.15: 89632930-89632930 on Build GRCh38
rs149221245 Chr.15: 89632836-89632836 on Build GRCh38
rs142786336 Chr.15: 89632900-89632900 on Build GRCh38
rs747755753 Chr.15: 89632942-89632942 on Build GRCh38
rs141463861 Chr.15: 89632854-89632854 on Build GRCh38
rs759963623 Chr.15: 89632841-89632841 on Build GRCh38
rs763169748 Chr.15: 89632931-89632931 on Build GRCh38
rs777302046 Chr.15: 89632964-89632964 on Build GRCh38
rs754318099 Chr.15: 89632840-89632840 on Build GRCh38
rs756146900 Chr.15: 89632866-89632866 on Build GRCh38
rs755180013 Chr.15: 89632800-89632800 on Build GRCh38
rs372753898 Chr.15: 89632913-89632913 on Build GRCh38
rs147069469 Chr.15: 89632868-89632868 on Build GRCh38
rs549295826 Chr.15: 89632813-89632813 on Build GRCh38
rs887506789 Chr.15: 89632904-89632904 on Build GRCh38
rs375079629 Chr.15: 89632887-89632887 on Build GRCh38
rs764430327 Chr.15: 89632980-89632980 on Build GRCh38
rs370197807 Chr.15: 89632792-89632792 on Build GRCh38
rs1002040263 Chr.15: 89632855-89632855 on Build GRCh38
rs763217993 Chr.15: 89632978-89632978 on Build GRCh38
rs148311253 Chr.15: 89632844-89632844 on Build GRCh38
rs777277054 Chr.15: 89632955-89632955 on Build GRCh38
rs749077709 Chr.15: 89632826-89632826 on Build GRCh38
rs760016965 Chr.15: 89632903-89632903 on Build GRCh38
rs745772459 Chr.15: 89632815-89632815 on Build GRCh38
rs767533531 Chr.15: 89632988-89632988 on Build GRCh38

View Full Product Details