Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 140537333-140537602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD2
Entrez Gene ID
255520
Gene Name
ELMO domain containing 2
Gene Aliases
9830169G11Rik
UniGene
Hs.450105
Gene Chromosome Location
Chr.4: 140524146-140553770 on Build GRCh38
Transcripts
NM_153702.3 XM_005262885.3 XM_005262886.2 XM_011531818.2 XM_011531819.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6866966 c.297A>G p.L99L Chr.4: 140537439-140537439 on Build GRCh38
1205212 c.287A>C p.K96T Chr.4: 140537429-140537429 on Build GRCh38
4394177 c.396G>A p.M132I Chr.4: 140537538-140537538 on Build GRCh38
6099271 c.341G>A p.S114N Chr.4: 140537483-140537483 on Build GRCh38
1051454 c.360T>C p.Y120Y Chr.4: 140537502-140537502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754716139 Chr.4: 140537466-140537466 on Build GRCh38
rs769738337 Chr.4: 140537481-140537481 on Build GRCh38
rs369960970 Chr.4: 140537486-140537486 on Build GRCh38
rs189023174 Chr.4: 140537509-140537509 on Build GRCh38
rs529218126 Chr.4: 140537505-140537505 on Build GRCh38
rs757512643 Chr.4: 140537437-140537437 on Build GRCh38
rs748000172 Chr.4: 140537563-140537563 on Build GRCh38
rs553898081 Chr.4: 140537517-140537517 on Build GRCh38
rs746809597 Chr.4: 140537374-140537374 on Build GRCh38
rs138133721 Chr.4: 140537499-140537499 on Build GRCh38
rs1047961619 Chr.4: 140537568-140537568 on Build GRCh38
rs764726417 Chr.4: 140537520-140537520 on Build GRCh38
rs769290006 Chr.4: 140537399-140537399 on Build GRCh38
rs761868502 Chr.4: 140537398-140537398 on Build GRCh38
rs770471853 Chr.4: 140537498-140537498 on Build GRCh38
rs776410145 Chr.4: 140537396-140537396 on Build GRCh38
rs762536633 Chr.4: 140537405-140537405 on Build GRCh38
rs868787800 Chr.4: 140537539-140537539 on Build GRCh38
rs879943796 Chr.4: 140537536-140537536 on Build GRCh38
rs777568889 Chr.4: 140537458-140537461 on Build GRCh38
rs577904155 Chr.4: 140537448-140537448 on Build GRCh38
rs756008600 Chr.4: 140537566-140537566 on Build GRCh38
rs112201741 Chr.4: 140537476-140537476 on Build GRCh38
rs772715383 Chr.4: 140537404-140537404 on Build GRCh38
rs556846456 Chr.4: 140537424-140537424 on Build GRCh38
rs746150538 Chr.4: 140537462-140537462 on Build GRCh38
rs772893341 Chr.4: 140537485-140537485 on Build GRCh38
rs750606475 Chr.4: 140537547-140537547 on Build GRCh38
rs753986525 Chr.4: 140537433-140537433 on Build GRCh38
rs377541996 Chr.4: 140537423-140537423 on Build GRCh38
rs149594258 Chr.4: 140537545-140537545 on Build GRCh38
rs767978161 Chr.4: 140537422-140537422 on Build GRCh38
rs765470660 Chr.4: 140537546-140537546 on Build GRCh38
rs1017675032 Chr.4: 140537490-140537490 on Build GRCh38
rs758651753 Chr.4: 140537548-140537548 on Build GRCh38
rs146977716 Chr.4: 140537493-140537493 on Build GRCh38
rs538889745 Chr.4: 140537510-140537510 on Build GRCh38
rs913104623 Chr.4: 140537367-140537367 on Build GRCh38
rs764209296 Chr.4: 140537429-140537429 on Build GRCh38
rs781109833 Chr.4: 140537467-140537467 on Build GRCh38
rs754435022 Chr.4: 140537537-140537537 on Build GRCh38
rs1024429640 Chr.4: 140537380-140537380 on Build GRCh38
rs780794601 Chr.4: 140537553-140537553 on Build GRCh38
rs751883088 Chr.4: 140537417-140537417 on Build GRCh38
rs768536262 Chr.4: 140537393-140537393 on Build GRCh38
rs567221180 Chr.4: 140537419-140537419 on Build GRCh38
rs141263113 Chr.4: 140537414-140537414 on Build GRCh38

View Full Product Details