Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44153184-44153447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCK
Entrez Gene ID
2645
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
UniGene
Hs.1270
Gene Chromosome Location
Chr.7: 44143213-44189439 on Build GRCh38
Transcripts
NM_000162.3 NM_033507.1 NM_033508.1 XM_017011966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6266548 c.84C>A p.D28E Chr.7: 44153422-44153422 on Build GRCh38
3638931 c.93G>A p.L31L Chr.7: 44153419-44153419 on Build GRCh38
4560586 c.78G>A p.E26E Chr.7: 44153428-44153428 on Build GRCh38
3881224 c.103C>T p.R35W Chr.7: 44153403-44153403 on Build GRCh38
4560096 c.82G>A p.E28K Chr.7: 44153430-44153430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs984651411 Chr.7: 44153410-44153410 on Build GRCh38
rs550111033 Chr.7: 44153371-44153371 on Build GRCh38
rs876661320 Chr.7: 44153420-44153420 on Build GRCh38
rs888594812 Chr.7: 44153293-44153293 on Build GRCh38
rs1053786524 Chr.7: 44153204-44153204 on Build GRCh38
rs564637857 Chr.7: 44153267-44153267 on Build GRCh38
rs373418736 Chr.7: 44153306-44153306 on Build GRCh38
rs779548342 Chr.7: 44153302-44153302 on Build GRCh38
rs747783371 Chr.7: 44153330-44153330 on Build GRCh38
rs193922288 Chr.7: 44153292-44153292 on Build GRCh38
rs769360432 Chr.7: 44153332-44153332 on Build GRCh38
rs942692580 Chr.7: 44153247-44153247 on Build GRCh38
rs749097393 Chr.7: 44153347-44153347 on Build GRCh38
rs868484983 Chr.7: 44153368-44153368 on Build GRCh38
rs780612692 Chr.7: 44153326-44153326 on Build GRCh38
rs886041690 Chr.7: 44153360-44153361 on Build GRCh38
rs794727236 Chr.7: 44153391-44153391 on Build GRCh38
rs764232985 Chr.7: 44153381-44153381 on Build GRCh38
rs759514960 Chr.7: 44153367-44153367 on Build GRCh38
rs775456214 Chr.7: 44153377-44153377 on Build GRCh38
rs377747439 Chr.7: 44153285-44153285 on Build GRCh38
rs193922279 Chr.7: 44153378-44153378 on Build GRCh38
rs878853246 Chr.7: 44153397-44153397 on Build GRCh38
rs193922287 Chr.7: 44153334-44153334 on Build GRCh38
rs193922270 Chr.7: 44153392-44153392 on Build GRCh38
rs754479025 Chr.7: 44153322-44153322 on Build GRCh38
rs193922259 Chr.7: 44153406-44153406 on Build GRCh38
rs1033196451 Chr.7: 44153228-44153228 on Build GRCh38
rs193922261 Chr.7: 44153402-44153402 on Build GRCh38
rs1005750049 Chr.7: 44153356-44153356 on Build GRCh38
rs545996860 Chr.7: 44153213-44153213 on Build GRCh38
rs774271708 Chr.7: 44153353-44153353 on Build GRCh38
rs754169218 Chr.7: 44153401-44153401 on Build GRCh38
rs911310769 Chr.7: 44153242-44153242 on Build GRCh38
rs144705480 Chr.7: 44153280-44153280 on Build GRCh38
rs866774967 Chr.7: 44153384-44153384 on Build GRCh38
rs759602984 Chr.7: 44153251-44153251 on Build GRCh38
rs762263694 Chr.7: 44153403-44153403 on Build GRCh38
rs199822205 Chr.7: 44153284-44153284 on Build GRCh38
rs77440690 Chr.7: 44153290-44153290 on Build GRCh38
rs760912915 Chr.7: 44153380-44153380 on Build GRCh38
rs746444094 Chr.7: 44153321-44153321 on Build GRCh38
rs267601516 Chr.7: 44153379-44153379 on Build GRCh38
rs193922286 Chr.7: 44153363-44153363 on Build GRCh38
rs377410513 Chr.7: 44153311-44153311 on Build GRCh38
rs919207783 Chr.7: 44153362-44153362 on Build GRCh38

View Full Product Details