The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005514.7 | NP_005505.2 | 6 - 7 | 1135 | 189 |
GenBank mRNA | AK124160.1 | - | 6 - 7 | 1318 | 189 |
AK129796.1 | - | 6 - 7 | 1069 | 189 | |
AK130166.1 | - | 6 - 7 | 1069 | 189 | |
AK130169.1 | - | 6 - 7 | 1075 | 189 | |
AK292226.1 | - | 6 - 7 | 1069 | 189 | |
AK300685.1 | - | 7 - 8 | 978 | 189 | |
AW273181.1 | - | 2 - 3 | 189 | ||
BC013828.2 | - | 2 - 3 | 153 | 189 | |
BC091497.1 | - | 6 - 7 | 1052 | 189 | |
D87665.1 | - | 7 - 8 | 1039 | 189 | |
L42345.1 | - | 6 - 7 | 1048 | 189 | |
M12678.1 | - | 6 - 7 | 1039 | 189 | |
M15470.1 | - | 6 - 7 | 1039 | 189 | |
M16102.1 | - | 6 - 7 | 189 | ||
M27540.1 | - | 5 - 6 | 862 | 189 | |
M28204.1 | - | 6 - 7 | 1012 | 189 | |
Z33453.1 | - | 6 - 7 | 1049 | 189 |