Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 167822951-167823176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY10
Entrez Gene ID
55811
Gene Name
adenylate cyclase 10, soluble
Gene Aliases
HCA2, HEL-S-7a, SAC, SACI, Sacy, hsAC
UniGene
Hs.320892
Gene Chromosome Location
Chr.1: 167809080-167915556 on Build GRCh38
Transcripts
NM_001167749.2 NM_001297772.1 NM_018417.5 XM_006711449.3 XM_011509760.2 XM_011509762.2 XM_011509763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354504 c.4058C>T p.P1353L Chr.1: 167823118-167823118 on Build GRCh38
7347452 c.4168+10G>T p.? Chr.1: 167822998-167822998 on Build GRCh38
4024823 c.4059G>A p.P1353P Chr.1: 167823117-167823117 on Build GRCh38
4493329 c.4134C>T p.F1378F Chr.1: 167823042-167823042 on Build GRCh38
899106 c.4162T>G p.Y1388D Chr.1: 167823014-167823014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767880875 Chr.1: 167823086-167823086 on Build GRCh38
rs372074672 Chr.1: 167823138-167823138 on Build GRCh38
rs764672173 Chr.1: 167823021-167823021 on Build GRCh38
rs369284643 Chr.1: 167823043-167823043 on Build GRCh38
rs202015026 Chr.1: 167822991-167822991 on Build GRCh38
rs145530045 Chr.1: 167823102-167823102 on Build GRCh38
rs760352710 Chr.1: 167822986-167822986 on Build GRCh38
rs767976390 Chr.1: 167822998-167822998 on Build GRCh38
rs201556412 Chr.1: 167823117-167823117 on Build GRCh38
rs776265715 Chr.1: 167822993-167822993 on Build GRCh38
rs200703036 Chr.1: 167823095-167823095 on Build GRCh38
rs771782930 Chr.1: 167822979-167822979 on Build GRCh38
rs904777660 Chr.1: 167823005-167823005 on Build GRCh38
rs775312673 Chr.1: 167822984-167822984 on Build GRCh38
rs752031595 Chr.1: 167823065-167823065 on Build GRCh38
rs772821053 Chr.1: 167823145-167823145 on Build GRCh38
rs996858935 Chr.1: 167823007-167823007 on Build GRCh38
rs866411878 Chr.1: 167823042-167823042 on Build GRCh38
rs376523925 Chr.1: 167823103-167823103 on Build GRCh38
rs946182273 Chr.1: 167822972-167822972 on Build GRCh38
rs148814511 Chr.1: 167823104-167823104 on Build GRCh38
rs200816878 Chr.1: 167823121-167823121 on Build GRCh38
rs1014646625 Chr.1: 167823113-167823113 on Build GRCh38
rs778997686 Chr.1: 167822975-167822975 on Build GRCh38
rs532854612 Chr.1: 167823116-167823116 on Build GRCh38
rs757424656 Chr.1: 167823109-167823109 on Build GRCh38
rs761409401 Chr.1: 167823142-167823142 on Build GRCh38
rs779830099 Chr.1: 167823120-167823120 on Build GRCh38
rs766688731 Chr.1: 167823094-167823094 on Build GRCh38
rs72697797 Chr.1: 167823118-167823118 on Build GRCh38
rs768377891 Chr.1: 167823123-167823123 on Build GRCh38
rs985519245 Chr.1: 167822996-167822996 on Build GRCh38
rs73039538 Chr.1: 167823101-167823101 on Build GRCh38
rs941572260 Chr.1: 167823062-167823062 on Build GRCh38
rs769327075 Chr.1: 167823143-167823143 on Build GRCh38
rs911299858 Chr.1: 167823016-167823016 on Build GRCh38
rs760049390 Chr.1: 167823083-167823083 on Build GRCh38
rs371824241 Chr.1: 167823029-167823029 on Build GRCh38
rs955338189 Chr.1: 167822992-167822992 on Build GRCh38
rs879030742 Chr.1: 167822999-167822999 on Build GRCh38
rs745759923 Chr.1: 167822976-167822976 on Build GRCh38

View Full Product Details