The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083913.1 | NP_001077382.1 | 3 - 4 | 432 | 102 |
NM_017787.4 | NP_060257.4 | 3 - 4 | 545 | 102 | |
XM_011539913.2 | XP_011538215.1 | 3 - 4 | 336 | 102 | |
GenBank mRNA | AK056285.1 | - | 3 - 4 | 429 | 102 |
AK300871.1 | - | 3 - 4 | 477 | 102 | |
AK316452.1 | - | 3 - 4 | 444 | 102 | |
AY656171.1 | - | 3 - 4 | 429 | 102 | |
BC113857.1 | - | 3 - 4 | 409 | 102 | |
BC114465.1 | - | 3 - 4 | 409 | 102 | |
CX163170.1 | - | 3 - 4 | 378 | 102 |