The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178311.2 | NP_842563.1 | 2 - 3 | 375 | 160 |
NM_178312.2 | NP_842564.1 | 2 - 3 | 303 | 160 | |
XM_005260863.4 | XP_005260920.1 | 2 - 3 | 730 | 160 | |
XM_005260864.4 | XP_005260921.1 | 3 - 4 | 883 | 160 | |
XM_005260865.4 | XP_005260922.1 | 3 - 4 | 883 | 160 | |
XM_011529391.2 | XP_011527693.1 | 2 - 3 | 297 | 160 | |
XM_011529392.2 | XP_011527694.1 | 3 - 4 | 822 | 160 | |
XM_011529393.2 | XP_011527695.1 | 2 - 3 | 669 | 160 | |
XM_017028126.1 | XP_016883615.1 | 2 - 3 | 292 | 160 | |
GenBank mRNA | BC040904.1 | - | 2 - 3 | 375 | 160 |
L20491.1 | - | 2 - 3 | 303 | 160 | |
L20492.1 | - | 1 - 2 | 353 | 160 |