Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112376221-112376494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL5
Entrez Gene ID
51703
Gene Name
acyl-CoA synthetase long-chain family member 5
Gene Aliases
ACS2, ACS5, FACL5
UniGene
Hs.11638
Gene Chromosome Location
Chr.10: 112374158-112428380 on Build GRCh38
Transcripts
NM_016234.3 NM_203379.1 NM_203380.1 XM_017016324.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3414688 c.28T>A p.W10R Chr.10: 112376337-112376337 on Build GRCh38
1745621 c.134C>T p.S45L Chr.10: 112376443-112376443 on Build GRCh38
3434131 c.100C>T p.H34Y Chr.10: 112376409-112376409 on Build GRCh38
2053142 c.27C>A p.L9L Chr.10: 112376336-112376336 on Build GRCh38
3434130 c.91G>A p.G31R Chr.10: 112376400-112376400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371363335 Chr.10: 112376375-112376375 on Build GRCh38
rs200599528 Chr.10: 112376374-112376374 on Build GRCh38
rs541260351 Chr.10: 112376437-112376437 on Build GRCh38
rs373249667 Chr.10: 112376455-112376455 on Build GRCh38
rs761080870 Chr.10: 112376317-112376317 on Build GRCh38
rs147992416 Chr.10: 112376444-112376444 on Build GRCh38
rs1041787659 Chr.10: 112376253-112376253 on Build GRCh38
rs758580630 Chr.10: 112376352-112376352 on Build GRCh38
rs1033824700 Chr.10: 112376363-112376363 on Build GRCh38
rs772895424 Chr.10: 112376398-112376398 on Build GRCh38
rs748801193 Chr.10: 112376307-112376307 on Build GRCh38
rs373993792 Chr.10: 112376349-112376349 on Build GRCh38
rs538435817 Chr.10: 112376256-112376256 on Build GRCh38
rs780125686 Chr.10: 112376372-112376372 on Build GRCh38
rs141233320 Chr.10: 112376434-112376434 on Build GRCh38
rs765690950 Chr.10: 112376313-112376313 on Build GRCh38
rs372279985 Chr.10: 112376412-112376412 on Build GRCh38
rs754412834 Chr.10: 112376345-112376345 on Build GRCh38
rs201482515 Chr.10: 112376415-112376415 on Build GRCh38
rs778105731 Chr.10: 112376353-112376353 on Build GRCh38
rs146797148 Chr.10: 112376396-112376396 on Build GRCh38
rs146947127 Chr.10: 112376438-112376438 on Build GRCh38
rs777390193 Chr.10: 112376473-112376474 on Build GRCh38
rs773994902 Chr.10: 112376309-112376309 on Build GRCh38
rs1000171729 Chr.10: 112376258-112376258 on Build GRCh38
rs202157884 Chr.10: 112376286-112376286 on Build GRCh38
rs752898656 Chr.10: 112376351-112376351 on Build GRCh38
rs766842660 Chr.10: 112376344-112376344 on Build GRCh38
rs866745436 Chr.10: 112376370-112376370 on Build GRCh38
rs199838168 Chr.10: 112376316-112376316 on Build GRCh38
rs750211609 Chr.10: 112376362-112376362 on Build GRCh38
rs754890178 Chr.10: 112376446-112376446 on Build GRCh38
rs370546224 Chr.10: 112376298-112376298 on Build GRCh38
rs149200937 Chr.10: 112376364-112376364 on Build GRCh38
rs903288191 Chr.10: 112376257-112376257 on Build GRCh38
rs758317937 Chr.10: 112376436-112376437 on Build GRCh38
rs753894860 Chr.10: 112376264-112376264 on Build GRCh38
rs752922649 Chr.10: 112376295-112376295 on Build GRCh38
rs375111541 Chr.10: 112376435-112376435 on Build GRCh38
rs375227875 Chr.10: 112376463-112376463 on Build GRCh38
rs752698749 Chr.10: 112376429-112376430 on Build GRCh38
rs747584394 Chr.10: 112376462-112376462 on Build GRCh38
rs145395373 Chr.10: 112376315-112376315 on Build GRCh38
rs993340712 Chr.10: 112376465-112376465 on Build GRCh38
rs375967711 Chr.10: 112376399-112376399 on Build GRCh38
rs536360557 Chr.10: 112376348-112376348 on Build GRCh38
rs755093138 Chr.10: 112376278-112376278 on Build GRCh38
rs778313797 Chr.10: 112376377-112376377 on Build GRCh38
rs777394378 Chr.10: 112376468-112376468 on Build GRCh38
rs768105743 Chr.10: 112376308-112376308 on Build GRCh38
rs1033041168 Chr.10: 112376265-112376265 on Build GRCh38
rs764405309 Chr.10: 112376436-112376436 on Build GRCh38
rs779192465 Chr.10: 112376285-112376285 on Build GRCh38
rs757020279 Chr.10: 112376296-112376296 on Build GRCh38
rs759966200 Chr.10: 112376432-112376432 on Build GRCh38
rs574547795 Chr.10: 112376443-112376443 on Build GRCh38
rs139217888 Chr.10: 112376433-112376433 on Build GRCh38

View Full Product Details