Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184186638-184186903 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF3
Entrez Gene ID
55324
Gene Name
ATP binding cassette subfamily F member 3
Gene Aliases
EST201864
UniGene
Hs.361323
Gene Chromosome Location
Chr.3: 184186075-184194012 on Build GRCh38
Transcripts
NM_018358.2 XM_011512959.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6256641 c.248G>T p.S83I Chr.3: 184186822-184186822 on Build GRCh38
1421285 c.206A>G p.Y69C Chr.3: 184186639-184186639 on Build GRCh38
5005905 c.224C>T p.A75V Chr.3: 184186798-184186798 on Build GRCh38
4540963 c.291G>A p.T97T Chr.3: 184186865-184186865 on Build GRCh38
729580 c.274C>T p.Q92* Chr.3: 184186848-184186848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748272932 Chr.3: 184186874-184186874 on Build GRCh38
rs55940723 Chr.3: 184186732-184186732 on Build GRCh38
rs562646826 Chr.3: 184186755-184186755 on Build GRCh38
rs767141305 Chr.3: 184186838-184186838 on Build GRCh38
rs759058672 Chr.3: 184186829-184186829 on Build GRCh38
rs772153210 Chr.3: 184186673-184186673 on Build GRCh38
rs144267642 Chr.3: 184186839-184186839 on Build GRCh38
rs763414518 Chr.3: 184186804-184186804 on Build GRCh38
rs754276906 Chr.3: 184186844-184186844 on Build GRCh38
rs755007073 Chr.3: 184186868-184186868 on Build GRCh38
rs768785470 Chr.3: 184186783-184186783 on Build GRCh38
rs1040042943 Chr.3: 184186713-184186714 on Build GRCh38
rs891963134 Chr.3: 184186796-184186796 on Build GRCh38
rs111599594 Chr.3: 184186668-184186668 on Build GRCh38
rs375560604 Chr.3: 184186685-184186685 on Build GRCh38
rs752411037 Chr.3: 184186768-184186768 on Build GRCh38
rs761019397 Chr.3: 184186748-184186749 on Build GRCh38
rs781268248 Chr.3: 184186871-184186871 on Build GRCh38
rs980076560 Chr.3: 184186847-184186847 on Build GRCh38
rs970813133 Chr.3: 184186727-184186727 on Build GRCh38
rs765587745 Chr.3: 184186694-184186694 on Build GRCh38
rs374230238 Chr.3: 184186787-184186787 on Build GRCh38
rs755854994 Chr.3: 184186840-184186840 on Build GRCh38
rs775055752 Chr.3: 184186810-184186810 on Build GRCh38
rs57210612 Chr.3: 184186720-184186720 on Build GRCh38
rs753548316 Chr.3: 184186845-184186845 on Build GRCh38
rs767264225 Chr.3: 184186763-184186763 on Build GRCh38
rs757145557 Chr.3: 184186848-184186848 on Build GRCh38
rs940390140 Chr.3: 184186703-184186703 on Build GRCh38
rs764577562 Chr.3: 184186767-184186768 on Build GRCh38
rs754600322 Chr.3: 184186761-184186761 on Build GRCh38
rs749187013 Chr.3: 184186669-184186669 on Build GRCh38
rs1010857764 Chr.3: 184186797-184186797 on Build GRCh38
rs758743610 Chr.3: 184186742-184186742 on Build GRCh38
rs776910470 Chr.3: 184186692-184186692 on Build GRCh38
rs778706360 Chr.3: 184186857-184186857 on Build GRCh38
rs199591659 Chr.3: 184186793-184186793 on Build GRCh38
rs748397306 Chr.3: 184186789-184186789 on Build GRCh38
rs778952224 Chr.3: 184186782-184186782 on Build GRCh38
rs142678058 Chr.3: 184186666-184186666 on Build GRCh38
rs751000127 Chr.3: 184186757-184186757 on Build GRCh38
rs763390482 Chr.3: 184186754-184186754 on Build GRCh38
rs775456294 Chr.3: 184186678-184186678 on Build GRCh38
rs773424973 Chr.3: 184186798-184186798 on Build GRCh38
rs745822416 Chr.3: 184186864-184186864 on Build GRCh38
rs1050283263 Chr.3: 184186765-184186765 on Build GRCh38
rs766789760 Chr.3: 184186809-184186809 on Build GRCh38
rs1004426132 Chr.3: 184186686-184186686 on Build GRCh38
rs368634729 Chr.3: 184186693-184186693 on Build GRCh38
rs372609098 Chr.3: 184186772-184186772 on Build GRCh38
rs763857323 Chr.3: 184186842-184186842 on Build GRCh38
rs749202656 Chr.3: 184186774-184186774 on Build GRCh38
rs777460496 Chr.3: 184186773-184186773 on Build GRCh38
rs773724236 Chr.3: 184186695-184186695 on Build GRCh38
rs1033558204 Chr.3: 184186792-184186792 on Build GRCh38
rs757121518 Chr.3: 184186775-184186775 on Build GRCh38

View Full Product Details