Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111727913-111728186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD10
Entrez Gene ID
80724
Gene Name
acyl-CoA dehydrogenase family member 10
Gene Aliases
-
UniGene
Hs.331141
Gene Chromosome Location
Chr.12: 111686053-111757107 on Build GRCh38
Transcripts
NM_001136538.1 NM_025247.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5970515 c.1116A>C p.K372N Chr.12: 111728016-111728016 on Build GRCh38
4593586 c.1118A>C p.D373A Chr.12: 111728018-111728018 on Build GRCh38
4146611 c.1233G>C p.E411D Chr.12: 111728040-111728040 on Build GRCh38
6591574 c.1071C>T p.G357G Chr.12: 111727971-111727971 on Build GRCh38
6591573 c.1071C>T p.G357G Chr.12: 111727971-111727971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975664219 Chr.12: 111728047-111728047 on Build GRCh38
rs11066013 Chr.12: 111728107-111728107 on Build GRCh38
rs762031537 Chr.12: 111727942-111727942 on Build GRCh38
rs772343029 Chr.12: 111728025-111728025 on Build GRCh38
rs1000650410 Chr.12: 111728019-111728019 on Build GRCh38
rs758721322 Chr.12: 111728092-111728092 on Build GRCh38
rs751451380 Chr.12: 111728066-111728066 on Build GRCh38
rs149558852 Chr.12: 111727980-111727980 on Build GRCh38
rs375092571 Chr.12: 111728068-111728068 on Build GRCh38
rs542306618 Chr.12: 111728099-111728099 on Build GRCh38
rs530140051 Chr.12: 111728059-111728059 on Build GRCh38
rs777516851 Chr.12: 111728089-111728089 on Build GRCh38
rs762763032 Chr.12: 111728054-111728054 on Build GRCh38
rs746211373 Chr.12: 111728023-111728023 on Build GRCh38
rs35358426 Chr.12: 111728052-111728052 on Build GRCh38
rs373192095 Chr.12: 111728021-111728021 on Build GRCh38
rs769037433 Chr.12: 111728110-111728110 on Build GRCh38
rs771000015 Chr.12: 111728003-111728004 on Build GRCh38
rs776764785 Chr.12: 111727941-111727941 on Build GRCh38
rs941483600 Chr.12: 111728060-111728060 on Build GRCh38
rs774510931 Chr.12: 111728095-111728096 on Build GRCh38
rs369542306 Chr.12: 111728132-111728132 on Build GRCh38
rs776675002 Chr.12: 111728042-111728042 on Build GRCh38
rs780573782 Chr.12: 111727987-111727987 on Build GRCh38
rs143739010 Chr.12: 111728137-111728137 on Build GRCh38
rs1038365580 Chr.12: 111728090-111728090 on Build GRCh38
rs138876094 Chr.12: 111728040-111728041 on Build GRCh38
rs923318197 Chr.12: 111727952-111727952 on Build GRCh38
rs925659687 Chr.12: 111728013-111728013 on Build GRCh38
rs768720282 Chr.12: 111728040-111728040 on Build GRCh38
rs776024275 Chr.12: 111728026-111728026 on Build GRCh38
rs373424367 Chr.12: 111728159-111728159 on Build GRCh38
rs767554445 Chr.12: 111727963-111727963 on Build GRCh38
rs754474740 Chr.12: 111727973-111727973 on Build GRCh38
rs747745769 Chr.12: 111728005-111728005 on Build GRCh38
rs895747686 Chr.12: 111728162-111728162 on Build GRCh38
rs752253650 Chr.12: 111728071-111728071 on Build GRCh38
rs755727414 Chr.12: 111728087-111728087 on Build GRCh38
rs978410826 Chr.12: 111728003-111728003 on Build GRCh38
rs780292003 Chr.12: 111728098-111728098 on Build GRCh38
rs967540247 Chr.12: 111727967-111727967 on Build GRCh38
rs908055923 Chr.12: 111728101-111728101 on Build GRCh38
rs541455128 Chr.12: 111728051-111728051 on Build GRCh38
rs557210190 Chr.12: 111727982-111727982 on Build GRCh38
rs962267406 Chr.12: 111728108-111728108 on Build GRCh38
rs200607092 Chr.12: 111728015-111728015 on Build GRCh38
rs749480203 Chr.12: 111728069-111728069 on Build GRCh38
rs138096024 Chr.12: 111727971-111727971 on Build GRCh38
rs758232313 Chr.12: 111727958-111727958 on Build GRCh38
rs773475147 Chr.12: 111728053-111728053 on Build GRCh38
rs749530509 Chr.12: 111727993-111727993 on Build GRCh38
rs747474122 Chr.12: 111728029-111728029 on Build GRCh38
rs368138546 Chr.12: 111727953-111727953 on Build GRCh38
rs765498227 Chr.12: 111727947-111727947 on Build GRCh38
rs747460658 Chr.12: 111728104-111728104 on Build GRCh38
rs777637232 Chr.12: 111728018-111728018 on Build GRCh38
rs773387270 Chr.12: 111728160-111728160 on Build GRCh38

View Full Product Details