Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC40
Entrez Gene ID
55036
Gene Name
coiled-coil domain containing 40
Gene Aliases
CILD15, FAP172
Chromosome Location
Chr.17: 80036632 - 80100613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243342.1 NP_001230271.1 7 - 8 1191 89
NM_017950.3 NP_060420.2 7 - 8 1191 89
XM_005257492.4 XP_005257549.1 7 - 8 1191 89
XM_011524963.2 XP_011523265.1 7 - 8 1228 89
XM_011524965.2 XP_011523267.1 7 - 8 1191 89
XM_017024807.1 XP_016880296.1 7 - 8 1191 89
GenBank mRNA AB046860.1 - 3 - 4 697 89
AK000760.1 - 7 - 8 1171 89
BC035251.1 - 7 - 8 1184 89
BC058288.1 - 7 - 8 1169 89

View Full Product Details