Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78767982-78768249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS7
Entrez Gene ID
11173
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 7
Gene Aliases
ADAM-TS 7, ADAM-TS7, ADAMTS-7
UniGene
Hs.16441
Gene Chromosome Location
Chr.15: 78759203-78811464 on Build GRCh38
Transcripts
NM_014272.4 XM_005254137.3 XM_011521166.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1283280 c.2591T>C p.L864P Chr.15: 78768187-78768187 on Build GRCh38
1374807 c.2590delC p.L864fs*93 Chr.15: 78768188-78768188 on Build GRCh38
5383885 c.2595C>T p.G865G Chr.15: 78768183-78768183 on Build GRCh38
1301476 c.2613G>T p.Q871H Chr.15: 78768165-78768165 on Build GRCh38
308922 c.2546T>A p.L849* Chr.15: 78768232-78768232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749798085 Chr.15: 78768137-78768137 on Build GRCh38
rs750549948 Chr.15: 78768116-78768116 on Build GRCh38
rs565309663 Chr.15: 78768213-78768213 on Build GRCh38
rs201688179 Chr.15: 78768207-78768207 on Build GRCh38
rs866994906 Chr.15: 78768053-78768053 on Build GRCh38
rs769436545 Chr.15: 78768181-78768181 on Build GRCh38
rs763002735 Chr.15: 78768190-78768190 on Build GRCh38
rs111276676 Chr.15: 78768089-78768089 on Build GRCh38
rs770667014 Chr.15: 78768163-78768163 on Build GRCh38
rs372729088 Chr.15: 78768196-78768196 on Build GRCh38
rs780067456 Chr.15: 78768128-78768128 on Build GRCh38
rs781236576 Chr.15: 78768094-78768094 on Build GRCh38
rs920196442 Chr.15: 78768100-78768100 on Build GRCh38
rs780167355 Chr.15: 78768212-78768212 on Build GRCh38
rs867460783 Chr.15: 78768033-78768033 on Build GRCh38
rs556389357 Chr.15: 78768111-78768112 on Build GRCh38
rs549286782 Chr.15: 78768095-78768096 on Build GRCh38
rs767250980 Chr.15: 78768206-78768206 on Build GRCh38
rs576824978 Chr.15: 78768215-78768215 on Build GRCh38
rs146487400 Chr.15: 78768182-78768182 on Build GRCh38
rs769031662 Chr.15: 78768147-78768147 on Build GRCh38
rs779402774 Chr.15: 78768152-78768152 on Build GRCh38
rs748430741 Chr.15: 78768153-78768153 on Build GRCh38
rs759227479 Chr.15: 78768177-78768177 on Build GRCh38
rs111612049 Chr.15: 78768065-78768065 on Build GRCh38
rs756207146 Chr.15: 78768125-78768125 on Build GRCh38
rs751406598 Chr.15: 78768192-78768192 on Build GRCh38
rs369211102 Chr.15: 78768130-78768130 on Build GRCh38
rs866735050 Chr.15: 78768132-78768132 on Build GRCh38
rs868394825 Chr.15: 78768048-78768048 on Build GRCh38
rs776449805 Chr.15: 78768166-78768166 on Build GRCh38
rs758723833 Chr.15: 78768227-78768227 on Build GRCh38
rs76539006 Chr.15: 78768226-78768226 on Build GRCh38
rs374389913 Chr.15: 78768048-78768049 on Build GRCh38
rs1019595651 Chr.15: 78768077-78768077 on Build GRCh38

View Full Product Details