The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198451.3 | NP_940853.1 | 1 - 1 | 1629 | 146 |
GenBank mRNA | BC012934.2 | - | 1 - 1 | 1643 | 146 |