Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT15
Entrez Gene ID
83849
Gene Name
synaptotagmin 15
Gene Aliases
CHR10SYT, sytXV
Chromosome Location
Chr.10: 46578434 - 46595170 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031912.4 NP_114118.2 7 - 8 1272 124
XM_017016750.1 XP_016872239.1 7 - 8 1282 124
GenBank mRNA AB109022.1 - 6 - 7 1119 124
AK127436.1 - 6 - 7 1866 124
AK290186.1 - 7 - 8 1186 124
AK293632.1 - 7 - 8 950 124

Gene Symbol
LOC102724488
Entrez Gene ID
102724488
Gene Name
synaptotagmin-15
Gene Aliases
-
Chromosome Location
Chr.10: 47745306 - 47763592 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006718093.3 XP_006718156.1 6 - 7 1866 124
XM_011540432.2 XP_011538734.1 6 - 7 1887 124
XM_017017020.1 XP_016872509.1 7 - 8 1282 124
XM_017017021.1 XP_016872510.1 7 - 8 1264 124
XM_017017024.1 XP_016872513.1 5 - 6 857 124

View Full Product Details