Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119895522-119895795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM30
Entrez Gene ID
11085
Gene Name
ADAM metallopeptidase domain 30
Gene Aliases
svph4
UniGene
Hs.283011
Gene Chromosome Location
Chr.1: 119893533-119896524 on Build GRCh38
Transcripts
NM_021794.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4595745 c.587C>T p.P196L Chr.1: 119895750-119895750 on Build GRCh38
169384 c.603C>T p.H201H Chr.1: 119895734-119895734 on Build GRCh38
1688426 c.799C>T p.P267S Chr.1: 119895538-119895538 on Build GRCh38
3399609 c.787C>T p.R263C Chr.1: 119895550-119895550 on Build GRCh38
6593701 c.789C>T p.R263R Chr.1: 119895548-119895548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201439972 Chr.1: 119895666-119895666 on Build GRCh38
rs140709609 Chr.1: 119895643-119895643 on Build GRCh38
rs375773692 Chr.1: 119895728-119895728 on Build GRCh38
rs773212860 Chr.1: 119895701-119895701 on Build GRCh38
rs966686307 Chr.1: 119895771-119895771 on Build GRCh38
rs1037530684 Chr.1: 119895661-119895661 on Build GRCh38
rs778427356 Chr.1: 119895763-119895763 on Build GRCh38
rs200088734 Chr.1: 119895550-119895550 on Build GRCh38
rs767986095 Chr.1: 119895739-119895739 on Build GRCh38
rs143465589 Chr.1: 119895707-119895707 on Build GRCh38
rs1031204471 Chr.1: 119895755-119895755 on Build GRCh38
rs758590430 Chr.1: 119895636-119895636 on Build GRCh38
rs868045361 Chr.1: 119895634-119895634 on Build GRCh38
rs776527983 Chr.1: 119895609-119895609 on Build GRCh38
rs779200633 Chr.1: 119895553-119895553 on Build GRCh38
rs757867438 Chr.1: 119895765-119895765 on Build GRCh38
rs771678408 Chr.1: 119895588-119895588 on Build GRCh38
rs1038778056 Chr.1: 119895601-119895601 on Build GRCh38
rs756630358 Chr.1: 119895759-119895759 on Build GRCh38
rs774253281 Chr.1: 119895744-119895744 on Build GRCh38
rs187002434 Chr.1: 119895679-119895679 on Build GRCh38
rs771687212 Chr.1: 119895667-119895667 on Build GRCh38
rs112374881 Chr.1: 119895650-119895650 on Build GRCh38
rs371062982 Chr.1: 119895547-119895547 on Build GRCh38
rs761726873 Chr.1: 119895689-119895689 on Build GRCh38
rs368586129 Chr.1: 119895549-119895549 on Build GRCh38
rs748631288 Chr.1: 119895655-119895655 on Build GRCh38
rs749907504 Chr.1: 119895764-119895764 on Build GRCh38
rs760391407 Chr.1: 119895593-119895593 on Build GRCh38
rs1024551020 Chr.1: 119895773-119895773 on Build GRCh38
rs376240673 Chr.1: 119895624-119895624 on Build GRCh38
rs753167602 Chr.1: 119895741-119895741 on Build GRCh38
rs999252293 Chr.1: 119895721-119895721 on Build GRCh38
rs587720609 Chr.1: 119895566-119895566 on Build GRCh38
rs775296308 Chr.1: 119895591-119895591 on Build GRCh38
rs774041463 Chr.1: 119895567-119895567 on Build GRCh38
rs749155212 Chr.1: 119895617-119895617 on Build GRCh38
rs781721317 Chr.1: 119895654-119895654 on Build GRCh38
rs41276634 Chr.1: 119895672-119895672 on Build GRCh38
rs886731921 Chr.1: 119895621-119895621 on Build GRCh38
rs766028098 Chr.1: 119895547-119895547 on Build GRCh38
rs371890150 Chr.1: 119895603-119895603 on Build GRCh38
rs776477537 Chr.1: 119895682-119895682 on Build GRCh38
rs367912978 Chr.1: 119895653-119895653 on Build GRCh38
rs770538963 Chr.1: 119895556-119895556 on Build GRCh38
rs756749170 Chr.1: 119895656-119895656 on Build GRCh38
rs587606101 Chr.1: 119895767-119895767 on Build GRCh38
rs745509046 Chr.1: 119895572-119895572 on Build GRCh38
rs761779917 Chr.1: 119895611-119895611 on Build GRCh38
rs1052426986 Chr.1: 119895614-119895614 on Build GRCh38
rs587699560 Chr.1: 119895694-119895694 on Build GRCh38
rs374434107 Chr.1: 119895548-119895548 on Build GRCh38
rs746769828 Chr.1: 119895678-119895678 on Build GRCh38
rs766517810 Chr.1: 119895638-119895638 on Build GRCh38
rs376536567 Chr.1: 119895675-119895675 on Build GRCh38
rs762977289 Chr.1: 119895703-119895703 on Build GRCh38

View Full Product Details