Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- GOLGA6L9
- Entrez Gene ID
-
440295
- Gene Name
- golgin A6 family-like 9
- Gene Aliases
-
GOLGA6L20
- Chromosome Location
-
Chr.15: 82429363 - 82439387 on Build GRCh38
- Gene Symbol
- GOLGA6L10
- Entrez Gene ID
-
647042
- Gene Name
- golgin A6 family-like 10
- Gene Aliases
-
GOLGA6L18
- Chromosome Location
-
Chr.15: 82339993 - 82349475 on Build GRCh38
- Gene Symbol
- GOLGA6L17P
- Entrez Gene ID
-
642402
- Gene Name
- golgin A6 family-like 17, pseudogene
- Gene Aliases
-
GOLGA6L21P
- Chromosome Location
-
Chr.15: 82519590 - 82526484 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111962.1 |
|
6 - 6 |
1501 |
125 |
GenBank mRNA |
AK301885.1 |
- |
- |
1627 |
125 |
- Gene Symbol
- GOLGA6L5P
- Entrez Gene ID
-
374650
- Gene Name
- golgin A6 family-like 5, pseudogene
- Gene Aliases
-
GOLGA6L5, GOLGA6L8
- Chromosome Location
-
Chr.15: 84506168 - 84516847 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK130562.1 |
- |
6 - 6 |
1158 |
125 |
View Full Product Details