Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNIH4
Entrez Gene ID
29097
Gene Name
cornichon family AMPA receptor auxiliary protein 4
Gene Aliases
CNIH-4, HSPC163
Chromosome Location
Chr.1: 224356811 - 224379452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277199.1 NP_001264128.1 3 - 4 645 96
NM_001277200.1 NP_001264129.1 3 - 4 367 96
NM_014184.3 NP_054903.1 3 - 4 367 96
NR_102347.1 2 - 3 348 96
GenBank mRNA AF161512.1 - 3 - 4 286 96
AK024569.1 - - 282 96
AK222634.1 - 3 - 4 272 96
AK312064.1 - 3 - 4 292 96
BC000573.1 - 3 - 4 282 96
BQ575976.1 - 2 - 3 96
DA185135.1 - 3 - 4 368 96
HY018269.1 - 2 - 3 264 96
HY145928.1 - 3 - 4 292 96

View Full Product Details