Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40395296-40395529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
UniGene
Hs.205457
Gene Chromosome Location
Chr.19: 40393762-40414718 on Build GRCh38
Transcripts
NM_020956.2 NM_181882.2 XM_011527171.2 XM_017027046.1 XM_017027047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6298349 c.2980C>A p.P994T Chr.19: 40395372-40395372 on Build GRCh38
5462848 c.2960C>A p.P987H Chr.19: 40395392-40395392 on Build GRCh38
3534056 c.2944G>A p.E982K Chr.19: 40395408-40395408 on Build GRCh38
3783240 c.2874G>A p.K958K Chr.19: 40395478-40395478 on Build GRCh38
4853945 c.3048C>T p.F1016F Chr.19: 40395304-40395304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748622767 Chr.19: 40395501-40395501 on Build GRCh38
rs762456536 Chr.19: 40395404-40395404 on Build GRCh38
rs548086012 Chr.19: 40395315-40395315 on Build GRCh38
rs562422065 Chr.19: 40395414-40395414 on Build GRCh38
rs142228732 Chr.19: 40395409-40395409 on Build GRCh38
rs758668778 Chr.19: 40395460-40395460 on Build GRCh38
rs104894714 Chr.19: 40395495-40395495 on Build GRCh38
rs996865233 Chr.19: 40395341-40395341 on Build GRCh38
rs770520776 Chr.19: 40395502-40395502 on Build GRCh38
rs778987729 Chr.19: 40395450-40395450 on Build GRCh38
rs759290682 Chr.19: 40395417-40395417 on Build GRCh38
rs764162630 Chr.19: 40395443-40395443 on Build GRCh38
rs1043055387 Chr.19: 40395379-40395379 on Build GRCh38
rs200424895 Chr.19: 40395395-40395395 on Build GRCh38
rs772591261 Chr.19: 40395400-40395400 on Build GRCh38
rs376423497 Chr.19: 40395453-40395453 on Build GRCh38
rs752590323 Chr.19: 40395429-40395429 on Build GRCh38
rs767386430 Chr.19: 40395419-40395419 on Build GRCh38
rs374750300 Chr.19: 40395441-40395441 on Build GRCh38
rs747451387 Chr.19: 40395396-40395396 on Build GRCh38
rs752638036 Chr.19: 40395339-40395339 on Build GRCh38
rs147842738 Chr.19: 40395504-40395504 on Build GRCh38
rs769263827 Chr.19: 40395399-40395399 on Build GRCh38
rs868592190 Chr.19: 40395377-40395377 on Build GRCh38
rs767439555 Chr.19: 40395316-40395316 on Build GRCh38
rs536876712 Chr.19: 40395384-40395384 on Build GRCh38
rs747471167 Chr.19: 40395492-40395492 on Build GRCh38
rs748965299 Chr.19: 40395411-40395431 on Build GRCh38
rs868460197 Chr.19: 40395459-40395459 on Build GRCh38
rs777695738 Chr.19: 40395355-40395355 on Build GRCh38
rs372961087 Chr.19: 40395447-40395447 on Build GRCh38
rs935218572 Chr.19: 40395393-40395393 on Build GRCh38
rs971346863 Chr.19: 40395432-40395432 on Build GRCh38
rs554688365 Chr.19: 40395394-40395394 on Build GRCh38
rs34766564 Chr.19: 40395338-40395338 on Build GRCh38
rs772380056 Chr.19: 40395390-40395390 on Build GRCh38
rs1027011237 Chr.19: 40395349-40395349 on Build GRCh38
rs879940695 Chr.19: 40395424-40395424 on Build GRCh38
rs566114248 Chr.19: 40395356-40395356 on Build GRCh38
rs776607887 Chr.19: 40395344-40395344 on Build GRCh38
rs1035734860 Chr.19: 40395477-40395477 on Build GRCh38
rs369688519 Chr.19: 40395493-40395493 on Build GRCh38
rs371192524 Chr.19: 40395405-40395405 on Build GRCh38
rs780382855 Chr.19: 40395483-40395483 on Build GRCh38
rs757620514 Chr.19: 40395350-40395350 on Build GRCh38
rs369224308 Chr.19: 40395444-40395444 on Build GRCh38
rs537040907 Chr.19: 40395507-40395507 on Build GRCh38
rs765924621 Chr.19: 40395353-40395353 on Build GRCh38
rs757419019 Chr.19: 40395360-40395360 on Build GRCh38
rs781470128 Chr.19: 40395393-40395394 on Build GRCh38
rs779042943 Chr.19: 40395361-40395361 on Build GRCh38
rs961710252 Chr.19: 40395466-40395466 on Build GRCh38
rs781679882 Chr.19: 40395496-40395496 on Build GRCh38

View Full Product Details