Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 243293100-243293351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDCCAG8
Entrez Gene ID
10806
Gene Name
serologically defined colon cancer antigen 8
Gene Aliases
BBS16, CCCAP, CCCAP SLSN7, HSPC085, NPHP10, NY-CO-8, SLSN7, hCCCAP
UniGene
Hs.591530
Gene Chromosome Location
Chr.1: 243255328-243500092 on Build GRCh38
Transcripts
NM_006642.3 XM_005273013.4 XM_005273018.1 XM_005273021.4 XM_005273022.3 XM_005273023.4 XM_006711727.3 XM_006711728.3 XM_011544021.2 XM_011544022.2 XM_011544023.2 XM_011544024.2 XM_011544026.2 XM_011544028.2 XM_011544029.2 XM_017000103.1 XM_017000104.1 XM_017000105.1 XM_017000106.1 XM_017000107.1 XM_017000108.1 XM_017000109.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3977367 c.557A>G p.H186R Chr.1: 243293101-243293101 on Build GRCh38
5846026 c.614C>T p.P205L Chr.1: 243293158-243293158 on Build GRCh38
6773700 c.674T>C p.L225P Chr.1: 243293218-243293218 on Build GRCh38
906595 c.654T>C p.S218S Chr.1: 243293198-243293198 on Build GRCh38
906596 c.659G>A p.G220D Chr.1: 243293203-243293203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769963982 Chr.1: 243293145-243293145 on Build GRCh38
rs756676848 Chr.1: 243293251-243293252 on Build GRCh38
rs771710977 Chr.1: 243293168-243293168 on Build GRCh38
rs968117633 Chr.1: 243293224-243293224 on Build GRCh38
rs190278956 Chr.1: 243293248-243293248 on Build GRCh38
rs752000188 Chr.1: 243293253-243293253 on Build GRCh38
rs780525378 Chr.1: 243293128-243293128 on Build GRCh38
rs1022080658 Chr.1: 243293220-243293220 on Build GRCh38
rs368972372 Chr.1: 243293303-243293303 on Build GRCh38
rs780216299 Chr.1: 243293251-243293251 on Build GRCh38
rs764042433 Chr.1: 243293187-243293187 on Build GRCh38
rs373999051 Chr.1: 243293205-243293205 on Build GRCh38
rs773304620 Chr.1: 243293163-243293163 on Build GRCh38
rs1012011593 Chr.1: 243293218-243293218 on Build GRCh38
rs760565857 Chr.1: 243293178-243293178 on Build GRCh38
rs552079971 Chr.1: 243293279-243293279 on Build GRCh38
rs761283265 Chr.1: 243293197-243293197 on Build GRCh38
rs748257473 Chr.1: 243293142-243293142 on Build GRCh38
rs755490466 Chr.1: 243293271-243293271 on Build GRCh38
rs900949463 Chr.1: 243293206-243293206 on Build GRCh38
rs747460316 Chr.1: 243293136-243293136 on Build GRCh38
rs923843519 Chr.1: 243293282-243293282 on Build GRCh38
rs750029026 Chr.1: 243293234-243293234 on Build GRCh38
rs763297129 Chr.1: 243293167-243293167 on Build GRCh38
rs143407309 Chr.1: 243293141-243293141 on Build GRCh38
rs775273268 Chr.1: 243293173-243293173 on Build GRCh38
rs951537520 Chr.1: 243293307-243293307 on Build GRCh38
rs753823148 Chr.1: 243293190-243293190 on Build GRCh38
rs758082302 Chr.1: 243293246-243293246 on Build GRCh38
rs771554391 Chr.1: 243293312-243293312 on Build GRCh38

View Full Product Details