Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15397493-15397748 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP8L
Entrez Gene ID
26993
Gene Name
A-kinase anchoring protein 8 like
Gene Aliases
HA95, HAP95, NAKAP, NAKAP95
UniGene
Hs.399800
Gene Chromosome Location
Chr.19: 15380048-15419121 on Build GRCh38
Transcripts
NM_001291478.1 NM_014371.3 NR_111971.1 XM_005259854.3 XM_011527916.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4750311 c.1353G>T p.E451D Chr.19: 15397572-15397572 on Build GRCh38
1304110 c.1323G>A p.K441K Chr.19: 15397602-15397602 on Build GRCh38
7318255 c.1380C>G p.I460M Chr.19: 15397545-15397545 on Build GRCh38
1391113 c.1336C>A p.L446I Chr.19: 15397589-15397589 on Build GRCh38
6244000 c.1328C>A p.T443K Chr.19: 15397597-15397597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780547911 Chr.19: 15397577-15397577 on Build GRCh38
rs368458552 Chr.19: 15397648-15397648 on Build GRCh38
rs776198134 Chr.19: 15397666-15397666 on Build GRCh38
rs1040730749 Chr.19: 15397519-15397519 on Build GRCh38
rs149593511 Chr.19: 15397647-15397647 on Build GRCh38
rs753836217 Chr.19: 15397717-15397717 on Build GRCh38
rs532848259 Chr.19: 15397586-15397586 on Build GRCh38
rs751377206 Chr.19: 15397624-15397624 on Build GRCh38
rs781614073 Chr.19: 15397547-15397547 on Build GRCh38
rs775439269 Chr.19: 15397619-15397619 on Build GRCh38
rs748495634 Chr.19: 15397656-15397656 on Build GRCh38
rs761636511 Chr.19: 15397694-15397694 on Build GRCh38
rs764115427 Chr.19: 15397690-15397690 on Build GRCh38
rs370686410 Chr.19: 15397638-15397638 on Build GRCh38
rs374419052 Chr.19: 15397611-15397611 on Build GRCh38
rs967005441 Chr.19: 15397654-15397654 on Build GRCh38
rs1032701620 Chr.19: 15397675-15397675 on Build GRCh38
rs762819421 Chr.19: 15397620-15397620 on Build GRCh38
rs757523394 Chr.19: 15397523-15397523 on Build GRCh38
rs202179580 Chr.19: 15397649-15397649 on Build GRCh38
rs1028832101 Chr.19: 15397605-15397605 on Build GRCh38
rs756325107 Chr.19: 15397704-15397704 on Build GRCh38
rs780024044 Chr.19: 15397643-15397643 on Build GRCh38
rs749001002 Chr.19: 15397546-15397546 on Build GRCh38
rs779190730 Chr.19: 15397696-15397696 on Build GRCh38
rs779443571 Chr.19: 15397655-15397655 on Build GRCh38
rs560088066 Chr.19: 15397689-15397689 on Build GRCh38
rs749776315 Chr.19: 15397578-15397578 on Build GRCh38
rs377346079 Chr.19: 15397632-15397632 on Build GRCh38
rs769298633 Chr.19: 15397684-15397684 on Build GRCh38
rs371764774 Chr.19: 15397691-15397691 on Build GRCh38
rs757177910 Chr.19: 15397629-15397629 on Build GRCh38
rs956595412 Chr.19: 15397661-15397661 on Build GRCh38
rs922607920 Chr.19: 15397543-15397543 on Build GRCh38
rs374802376 Chr.19: 15397710-15397710 on Build GRCh38
rs767851157 Chr.19: 15397631-15397631 on Build GRCh38
rs762203464 Chr.19: 15397593-15397593 on Build GRCh38
rs745520435 Chr.19: 15397682-15397682 on Build GRCh38
rs79796304 Chr.19: 15397729-15397729 on Build GRCh38
rs769835797 Chr.19: 15397557-15397557 on Build GRCh38
rs1047148251 Chr.19: 15397571-15397571 on Build GRCh38
rs2058322 Chr.19: 15397551-15397551 on Build GRCh38
rs762573190 Chr.19: 15397688-15397688 on Build GRCh38
rs772490640 Chr.19: 15397663-15397663 on Build GRCh38
rs750628449 Chr.19: 15397703-15397703 on Build GRCh38
rs769018908 Chr.19: 15397585-15397585 on Build GRCh38
rs764132259 Chr.19: 15397622-15397622 on Build GRCh38
rs770545443 Chr.19: 15397599-15397599 on Build GRCh38

View Full Product Details