The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178516.3 | NP_848611.2 | 11 - 12 | 1991 | 97 |
GenBank mRNA | AK092858.1 | - | 11 - 12 | 1883 | 97 |
AK092906.1 | - | 10 - 11 | 1629 | 97 | |
AK292004.1 | - | 11 - 12 | 1900 | 97 | |
AK300915.1 | - | 10 - 11 | 1650 | 97 | |
BC034287.1 | - | 8 - 9 | 1340 | 97 |