Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 60055809-60056070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANK3
Entrez Gene ID
288
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases
ANKYRIN-G, MRT37
UniGene
Hs.499725
Gene Chromosome Location
Chr.10: 60026298-60733526 on Build GRCh38
Transcripts
NM_001149.3 NM_001204403.1 NM_001204404.1 NM_001320874.1 NM_020987.4 XM_005269715.3 XM_006717796.3 XM_006717802.3 XM_011539708.2 XM_011539709.2 XM_017016110.1 XM_017016111.1 XM_017016112.1 XM_017016113.1 XM_017016114.1 XM_017016115.1 XM_017016116.1 XM_017016117.1 XM_017016118.1 XM_017016119.1 XM_017016120.1 XM_017016121.1 XM_017016122.1 XM_017016123.1 XM_017016124.1 XM_017016125.1 XM_017016126.1 XM_017016127.1 XM_017016128.1 XM_017016129.1 XM_017016130.1 XM_017016131.1 XM_017016132.1 XM_017016133.1 XM_017016134.1 XM_017016135.1 XM_017016136.1 XM_017016137.1 XM_017016138.1 XM_017016139.1 XM_017016140.1 XM_017016141.1 XM_017016142.1 XM_017016143.1 XM_017016144.1 XM_017016145.1 XM_017016146.1 XM_017016147.1 XM_017016148.1 XM_017016149.1 XM_017016150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599922 c.2758C>G p.P920A Chr.10: 60055837-60055837 on Build GRCh38
6599921 c.12886C>G p.P4296A Chr.10: 60055837-60055837 on Build GRCh38
427725 c.2575G>C p.D859H Chr.10: 60056020-60056020 on Build GRCh38
5370893 c.12817G>A p.D4273N Chr.10: 60055906-60055906 on Build GRCh38
4661597 c.12765A>C p.T4255T Chr.10: 60055958-60055958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs797045231 Chr.10: 60055971-60055971 on Build GRCh38
rs769573528 Chr.10: 60055960-60055960 on Build GRCh38
rs748955659 Chr.10: 60055959-60055959 on Build GRCh38
rs147941173 Chr.10: 60055995-60055995 on Build GRCh38
rs944249613 Chr.10: 60056010-60056010 on Build GRCh38
rs553218660 Chr.10: 60055886-60055886 on Build GRCh38
rs759965221 Chr.10: 60055998-60055998 on Build GRCh38
rs761730090 Chr.10: 60056027-60056027 on Build GRCh38
rs754817927 Chr.10: 60056033-60056033 on Build GRCh38
rs753206170 Chr.10: 60055885-60055885 on Build GRCh38
rs752702900 Chr.10: 60056043-60056043 on Build GRCh38
rs200666755 Chr.10: 60055951-60055951 on Build GRCh38
rs766720542 Chr.10: 60055847-60055847 on Build GRCh38
rs141051825 Chr.10: 60055856-60055856 on Build GRCh38
rs754730320 Chr.10: 60055930-60055930 on Build GRCh38
rs914801576 Chr.10: 60055887-60055887 on Build GRCh38
rs764325997 Chr.10: 60056025-60056025 on Build GRCh38
rs865944748 Chr.10: 60056000-60056000 on Build GRCh38
rs780934694 Chr.10: 60056034-60056034 on Build GRCh38
rs764237893 Chr.10: 60055889-60055889 on Build GRCh38
rs774766304 Chr.10: 60055854-60055854 on Build GRCh38
rs768167956 Chr.10: 60055869-60055869 on Build GRCh38
rs776176499 Chr.10: 60056009-60056009 on Build GRCh38
rs79971091 Chr.10: 60055974-60055974 on Build GRCh38
rs995936040 Chr.10: 60055983-60055983 on Build GRCh38
rs746822793 Chr.10: 60055908-60055908 on Build GRCh38
rs748059914 Chr.10: 60055941-60055941 on Build GRCh38
rs772904316 Chr.10: 60055846-60055846 on Build GRCh38
rs74961590 Chr.10: 60055907-60055907 on Build GRCh38
rs773811291 Chr.10: 60055961-60055961 on Build GRCh38
rs773627391 Chr.10: 60055926-60055926 on Build GRCh38
rs781133118 Chr.10: 60055937-60055937 on Build GRCh38
rs1039776718 Chr.10: 60055940-60055940 on Build GRCh38
rs780899852 Chr.10: 60055894-60055894 on Build GRCh38
rs368823652 Chr.10: 60056028-60056028 on Build GRCh38
rs750563582 Chr.10: 60056029-60056029 on Build GRCh38
rs772538751 Chr.10: 60056005-60056005 on Build GRCh38
rs12261793 Chr.10: 60055954-60055954 on Build GRCh38

View Full Product Details