Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 44808248-44808521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP8
Entrez Gene ID
23779
Gene Name
Rho GTPase activating protein 8
Gene Aliases
BPGAP1, PP610
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44752558-44862784 on Build GRCh38
Transcripts
NM_001017526.1 NM_001198726.1 NM_181335.2 NM_181334.5

Gene Symbol
PRR5-ARHGAP8
Entrez Gene ID
553158
Gene Name
PRR5-ARHGAP8 readthrough
Gene Aliases
-
UniGene
Hs.102336
Gene Chromosome Location
Chr.22: 44702198-44862784 on Build GRCh38
Transcripts
NM_001017526.1 NM_001198726.1 NM_181335.2 NM_181334.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4571769 c.169T>C p.Y57H Chr.22: 44808308-44808308 on Build GRCh38
3740466 c.293A>T p.D98V Chr.22: 44808432-44808432 on Build GRCh38
4518377 c.258_259CC>TT p.(=) Chr.22: 44808397-44808398 on Build GRCh38
3062770 c.407+4C>T p.? Chr.22: 44808442-44808442 on Build GRCh38
4972428 c.244C>T p.R82W Chr.22: 44808383-44808383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542654729 Chr.22: 44808414-44808414 on Build GRCh38
rs149715860 Chr.22: 44808325-44808325 on Build GRCh38
rs991091335 Chr.22: 44808353-44808353 on Build GRCh38
rs777071862 Chr.22: 44808423-44808423 on Build GRCh38
rs766698640 Chr.22: 44808365-44808365 on Build GRCh38
rs199881939 Chr.22: 44808474-44808474 on Build GRCh38
rs754599260 Chr.22: 44808427-44808427 on Build GRCh38
rs148849830 Chr.22: 44808356-44808356 on Build GRCh38
rs565633378 Chr.22: 44808343-44808343 on Build GRCh38
rs754686783 Chr.22: 44808490-44808490 on Build GRCh38
rs137919354 Chr.22: 44808364-44808364 on Build GRCh38
rs368556464 Chr.22: 44808331-44808331 on Build GRCh38
rs374300088 Chr.22: 44808442-44808442 on Build GRCh38
rs751900302 Chr.22: 44808368-44808368 on Build GRCh38
rs891197527 Chr.22: 44808322-44808322 on Build GRCh38
rs752542196 Chr.22: 44808301-44808301 on Build GRCh38
rs757999023 Chr.22: 44808476-44808477 on Build GRCh38
rs370461525 Chr.22: 44808464-44808464 on Build GRCh38
rs140846436 Chr.22: 44808319-44808319 on Build GRCh38
rs778613789 Chr.22: 44808461-44808472 on Build GRCh38
rs777570790 Chr.22: 44808312-44808312 on Build GRCh38
rs752361663 Chr.22: 44808464-44808464 on Build GRCh38
rs370106353 Chr.22: 44808289-44808289 on Build GRCh38
rs367903324 Chr.22: 44808448-44808448 on Build GRCh38
rs746593109 Chr.22: 44808482-44808482 on Build GRCh38
rs763757406 Chr.22: 44808451-44808451 on Build GRCh38
rs141930874 Chr.22: 44808377-44808377 on Build GRCh38
rs770528726 Chr.22: 44808340-44808340 on Build GRCh38
rs141907499 Chr.22: 44808355-44808355 on Build GRCh38
rs116110380 Chr.22: 44808384-44808384 on Build GRCh38
rs571482121 Chr.22: 44808370-44808370 on Build GRCh38
rs200183405 Chr.22: 44808415-44808415 on Build GRCh38
rs774418460 Chr.22: 44808449-44808449 on Build GRCh38
rs755997106 Chr.22: 44808304-44808304 on Build GRCh38
rs6007304 Chr.22: 44808383-44808383 on Build GRCh38
rs778417488 Chr.22: 44808326-44808326 on Build GRCh38
rs985852927 Chr.22: 44808461-44808461 on Build GRCh38
rs373035947 Chr.22: 44808295-44808295 on Build GRCh38
rs192449845 Chr.22: 44808283-44808283 on Build GRCh38
rs1049258921 Chr.22: 44808386-44808386 on Build GRCh38
rs768880228 Chr.22: 44808420-44808420 on Build GRCh38
rs959478163 Chr.22: 44808344-44808344 on Build GRCh38
rs771718778 Chr.22: 44808335-44808335 on Build GRCh38
rs902659548 Chr.22: 44808352-44808352 on Build GRCh38
rs763123710 Chr.22: 44808443-44808443 on Build GRCh38
rs112566913 Chr.22: 44808500-44808500 on Build GRCh38
rs878994473 Chr.22: 44808379-44808379 on Build GRCh38
rs750425496 Chr.22: 44808467-44808467 on Build GRCh38
rs146945151 Chr.22: 44808360-44808360 on Build GRCh38
rs888639412 Chr.22: 44808416-44808416 on Build GRCh38
rs557912750 Chr.22: 44808361-44808361 on Build GRCh38
rs538343031 Chr.22: 44808371-44808371 on Build GRCh38
rs371998862 Chr.22: 44808334-44808334 on Build GRCh38
rs200981266 Chr.22: 44808418-44808418 on Build GRCh38
rs757251755 Chr.22: 44808381-44808381 on Build GRCh38
rs866097893 Chr.22: 44808367-44808367 on Build GRCh38
rs369378992 Chr.22: 44808465-44808465 on Build GRCh38
rs748559516 Chr.22: 44808430-44808430 on Build GRCh38
rs759829638 Chr.22: 44808450-44808450 on Build GRCh38
rs757872203 Chr.22: 44808469-44808469 on Build GRCh38
rs148162583 Chr.22: 44808431-44808431 on Build GRCh38
rs565400046 Chr.22: 44808459-44808459 on Build GRCh38

View Full Product Details