Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 87945516-87945789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALC
Entrez Gene ID
2581
Gene Name
galactosylceramidase
Gene Aliases
-
UniGene
Hs.513439
Gene Chromosome Location
Chr.14: 87933014-87993665 on Build GRCh38
Transcripts
NM_000153.3 NM_001201401.1 NM_001201402.1 XM_011536618.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7272468 c.1649G>A p.S550N Chr.14: 87945574-87945574 on Build GRCh38
6668924 c.1335delT p.F445fs*52 Chr.14: 87945720-87945720 on Build GRCh38
5552324 c.1630G>A p.D544N Chr.14: 87945593-87945593 on Build GRCh38
7272469 c.1571G>A p.S524N Chr.14: 87945574-87945574 on Build GRCh38
6668922 c.1466C>T p.A489V Chr.14: 87945589-87945589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772686301 Chr.14: 87945585-87945585 on Build GRCh38
rs749893889 Chr.14: 87945632-87945632 on Build GRCh38
rs941906996 Chr.14: 87945752-87945752 on Build GRCh38
rs774239455 Chr.14: 87945619-87945619 on Build GRCh38
rs794727116 Chr.14: 87945680-87945680 on Build GRCh38
rs200378205 Chr.14: 87945631-87945631 on Build GRCh38
rs387906952 Chr.14: 87945593-87945593 on Build GRCh38
rs886050864 Chr.14: 87945581-87945581 on Build GRCh38
rs771079019 Chr.14: 87945596-87945596 on Build GRCh38
rs911390493 Chr.14: 87945547-87945547 on Build GRCh38
rs557464603 Chr.14: 87945604-87945604 on Build GRCh38
rs9672064 Chr.14: 87945591-87945591 on Build GRCh38
rs774001462 Chr.14: 87945713-87945713 on Build GRCh38
rs758827171 Chr.14: 87945657-87945657 on Build GRCh38
rs746062460 Chr.14: 87945690-87945690 on Build GRCh38
rs747006038 Chr.14: 87945670-87945670 on Build GRCh38
rs771025782 Chr.14: 87945678-87945678 on Build GRCh38
rs753164684 Chr.14: 87945651-87945651 on Build GRCh38
rs201055535 Chr.14: 87945594-87945594 on Build GRCh38
rs778349065 Chr.14: 87945661-87945661 on Build GRCh38
rs756543855 Chr.14: 87945602-87945603 on Build GRCh38
rs749597090 Chr.14: 87945552-87945552 on Build GRCh38
rs375867319 Chr.14: 87945682-87945682 on Build GRCh38
rs748573754 Chr.14: 87945566-87945566 on Build GRCh38
rs766016339 Chr.14: 87945758-87945758 on Build GRCh38
rs367327 Chr.14: 87945603-87945603 on Build GRCh38
rs967173797 Chr.14: 87945759-87945759 on Build GRCh38
rs761731174 Chr.14: 87945718-87945718 on Build GRCh38
rs776857280 Chr.14: 87945599-87945599 on Build GRCh38
rs779511828 Chr.14: 87945562-87945562 on Build GRCh38
rs753565491 Chr.14: 87945639-87945639 on Build GRCh38
rs755353417 Chr.14: 87945648-87945648 on Build GRCh38
rs760213309 Chr.14: 87945755-87945755 on Build GRCh38
rs770047605 Chr.14: 87945702-87945702 on Build GRCh38
rs370952794 Chr.14: 87945656-87945656 on Build GRCh38
rs771834033 Chr.14: 87945737-87945737 on Build GRCh38
rs200960659 Chr.14: 87945637-87945637 on Build GRCh38
rs146286491 Chr.14: 87945650-87945650 on Build GRCh38
rs751786254 Chr.14: 87945686-87945686 on Build GRCh38
rs755406507 Chr.14: 87945561-87945561 on Build GRCh38
rs761631708 Chr.14: 87945628-87945628 on Build GRCh38
rs558341768 Chr.14: 87945754-87945755 on Build GRCh38
rs773214484 Chr.14: 87945739-87945739 on Build GRCh38
rs752279674 Chr.14: 87945598-87945599 on Build GRCh38
rs1050027845 Chr.14: 87945543-87945543 on Build GRCh38
rs779915053 Chr.14: 87945545-87945545 on Build GRCh38
rs760730368 Chr.14: 87945688-87945688 on Build GRCh38
rs184748969 Chr.14: 87945636-87945636 on Build GRCh38

View Full Product Details