Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17327309-17327556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO8
Entrez Gene ID
57719
Gene Name
anoctamin 8
Gene Aliases
KIAA1623, TMEM16H
UniGene
Hs.590990
Gene Chromosome Location
Chr.19: 17323223-17334834 on Build GRCh38
Transcripts
NM_020959.2 XM_017027048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589672 c.2502C>T p.F834F Chr.19: 17327486-17327486 on Build GRCh38
3692479 c.2534C>T p.S845L Chr.19: 17327454-17327454 on Build GRCh38
6501429 c.2568C>T p.L856L Chr.19: 17327328-17327328 on Build GRCh38
4890418 c.2544G>T p.V848V Chr.19: 17327444-17327444 on Build GRCh38
4750515 c.2452G>A p.V818M Chr.19: 17327536-17327536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs527269675 Chr.19: 17327334-17327334 on Build GRCh38
rs777469846 Chr.19: 17327348-17327352 on Build GRCh38
rs768014396 Chr.19: 17327441-17327441 on Build GRCh38
rs956332627 Chr.19: 17327411-17327411 on Build GRCh38
rs765432336 Chr.19: 17327521-17327521 on Build GRCh38
rs748802558 Chr.19: 17327418-17327418 on Build GRCh38
rs569885331 Chr.19: 17327430-17327430 on Build GRCh38
rs761229392 Chr.19: 17327446-17327446 on Build GRCh38
rs957521726 Chr.19: 17327471-17327471 on Build GRCh38
rs920116771 Chr.19: 17327474-17327474 on Build GRCh38
rs549976404 Chr.19: 17327427-17327427 on Build GRCh38
rs567330096 Chr.19: 17327393-17327393 on Build GRCh38
rs1001222376 Chr.19: 17327371-17327371 on Build GRCh38
rs1030661026 Chr.19: 17327376-17327376 on Build GRCh38
rs753940125 Chr.19: 17327458-17327458 on Build GRCh38
rs773811139 Chr.19: 17327443-17327443 on Build GRCh38
rs755079048 Chr.19: 17327493-17327493 on Build GRCh38
rs752877827 Chr.19: 17327531-17327531 on Build GRCh38
rs1024693683 Chr.19: 17327424-17327424 on Build GRCh38
rs751604601 Chr.19: 17327486-17327486 on Build GRCh38
rs372002710 Chr.19: 17327401-17327401 on Build GRCh38
rs199668891 Chr.19: 17327477-17327477 on Build GRCh38
rs773944466 Chr.19: 17327429-17327429 on Build GRCh38
rs894528980 Chr.19: 17327390-17327390 on Build GRCh38
rs545935904 Chr.19: 17327384-17327384 on Build GRCh38
rs771023313 Chr.19: 17327413-17327415 on Build GRCh38
rs762849091 Chr.19: 17327341-17327341 on Build GRCh38
rs763978088 Chr.19: 17327383-17327383 on Build GRCh38
rs781051384 Chr.19: 17327400-17327400 on Build GRCh38
rs376028722 Chr.19: 17327454-17327454 on Build GRCh38
rs967980362 Chr.19: 17327352-17327352 on Build GRCh38
rs547221815 Chr.19: 17327379-17327379 on Build GRCh38
rs774950009 Chr.19: 17327417-17327417 on Build GRCh38
rs776517161 Chr.19: 17327423-17327423 on Build GRCh38
rs746912281 Chr.19: 17327412-17327413 on Build GRCh38
rs758498116 Chr.19: 17327359-17327359 on Build GRCh38

View Full Product Details