The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004096.4 | NP_004087.1 | 3 - 3 | 2348 | 155 |
GenBank mRNA | BC005057.1 | - | 3 - 3 | 2074 | 155 |
BC050633.1 | - | 3 - 3 | 2287 | 155 |