Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 43891956-43892227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH3
Entrez Gene ID
221120
Gene Name
alkB homolog 3, alpha-ketoglutaratedependent dioxygenase
Gene Aliases
ABH3, DEPC-1, DEPC1, PCA1, hABH3
UniGene
Hs.720708
Gene Chromosome Location
Chr.11: 43880807-43920275 on Build GRCh38
Transcripts
NM_139178.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3447301 c.376A>G p.T126A Chr.11: 43892046-43892046 on Build GRCh38
3979637 c.404G>A p.W135* Chr.11: 43892074-43892074 on Build GRCh38
5039387 c.400G>A p.A134T Chr.11: 43892070-43892070 on Build GRCh38
926917 c.459C>T p.H153H Chr.11: 43892129-43892129 on Build GRCh38
7245695 c.384G>A p.Q128Q Chr.11: 43892054-43892054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762180324 Chr.11: 43892112-43892112 on Build GRCh38
rs751160980 Chr.11: 43892049-43892049 on Build GRCh38
rs886508660 Chr.11: 43892081-43892081 on Build GRCh38
rs376360860 Chr.11: 43892166-43892166 on Build GRCh38
rs746764671 Chr.11: 43891999-43891999 on Build GRCh38
rs747276251 Chr.11: 43892078-43892078 on Build GRCh38
rs755901381 Chr.11: 43892155-43892155 on Build GRCh38
rs766048497 Chr.11: 43892048-43892048 on Build GRCh38
rs898367089 Chr.11: 43892097-43892097 on Build GRCh38
rs16937692 Chr.11: 43892054-43892054 on Build GRCh38
rs754689214 Chr.11: 43892138-43892138 on Build GRCh38
rs753864583 Chr.11: 43892167-43892167 on Build GRCh38
rs749914379 Chr.11: 43892033-43892033 on Build GRCh38
rs764037018 Chr.11: 43892037-43892037 on Build GRCh38
rs3740985 Chr.11: 43892157-43892157 on Build GRCh38
rs762294027 Chr.11: 43892022-43892022 on Build GRCh38
rs375859361 Chr.11: 43892012-43892012 on Build GRCh38
rs979409945 Chr.11: 43891998-43891998 on Build GRCh38
rs775691855 Chr.11: 43892076-43892076 on Build GRCh38
rs185371281 Chr.11: 43892099-43892099 on Build GRCh38
rs572226598 Chr.11: 43892190-43892190 on Build GRCh38
rs887411487 Chr.11: 43892125-43892125 on Build GRCh38
rs765921341 Chr.11: 43892132-43892132 on Build GRCh38
rs372213455 Chr.11: 43891990-43891990 on Build GRCh38
rs767103834 Chr.11: 43892143-43892143 on Build GRCh38
rs777193889 Chr.11: 43892108-43892108 on Build GRCh38
rs765760670 Chr.11: 43892032-43892032 on Build GRCh38
rs765666915 Chr.11: 43892114-43892114 on Build GRCh38
rs768997072 Chr.11: 43892091-43892091 on Build GRCh38
rs762390141 Chr.11: 43892130-43892130 on Build GRCh38
rs747708970 Chr.11: 43892185-43892185 on Build GRCh38
rs1017176346 Chr.11: 43892096-43892096 on Build GRCh38
rs751480702 Chr.11: 43892088-43892088 on Build GRCh38
rs772144965 Chr.11: 43892075-43892075 on Build GRCh38
rs780348729 Chr.11: 43892178-43892178 on Build GRCh38
rs777703982 Chr.11: 43892059-43892059 on Build GRCh38
rs969626961 Chr.11: 43892191-43892191 on Build GRCh38
rs751036010 Chr.11: 43892135-43892135 on Build GRCh38
rs756023889 Chr.11: 43892055-43892055 on Build GRCh38
rs145573011 Chr.11: 43892127-43892127 on Build GRCh38
rs757857296 Chr.11: 43892035-43892035 on Build GRCh38
rs764603732 Chr.11: 43892013-43892013 on Build GRCh38
rs745969101 Chr.11: 43892070-43892070 on Build GRCh38
rs777105819 Chr.11: 43892021-43892021 on Build GRCh38
rs757231033 Chr.11: 43892170-43892170 on Build GRCh38
rs772191496 Chr.11: 43892002-43892002 on Build GRCh38
rs776340849 Chr.11: 43892201-43892201 on Build GRCh38
rs780894901 Chr.11: 43892158-43892158 on Build GRCh38
rs944525950 Chr.11: 43892067-43892067 on Build GRCh38
rs752442550 Chr.11: 43892154-43892154 on Build GRCh38
rs754599411 Chr.11: 43892051-43892051 on Build GRCh38
rs757042976 Chr.11: 43892104-43892114 on Build GRCh38

View Full Product Details