The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006305.3 | NP_006296.1 | 7 - 7 | 1001 | 145 |
GenBank mRNA | AK223280.1 | - | 7 - 7 | 948 | 146 |
AY007110.1 | - | 5 - 5 | 539 | 145 | |
BC007200.1 | - | 7 - 7 | 963 | 145 |