Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18184962-18185207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH5
Entrez Gene ID
54890
Gene Name
alkB homolog 5, RNA demethylase
Gene Aliases
ABH5, OFOXD, OFOXD1
UniGene
Hs.744130
Gene Chromosome Location
Chr.17: 18183553-18209954 on Build GRCh38
Transcripts
NM_017758.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7259043 c.729G>A p.V243V Chr.17: 18184972-18184972 on Build GRCh38
4878438 c.745C>T p.R249C Chr.17: 18184988-18184988 on Build GRCh38
4507018 c.735C>T p.S245S Chr.17: 18184978-18184978 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760709418 Chr.17: 18185032-18185032 on Build GRCh38
rs1054361097 Chr.17: 18185184-18185184 on Build GRCh38
rs768763088 Chr.17: 18185039-18185040 on Build GRCh38
rs757073318 Chr.17: 18185180-18185180 on Build GRCh38
rs371096564 Chr.17: 18184985-18184985 on Build GRCh38
rs192322570 Chr.17: 18185131-18185131 on Build GRCh38
rs868094334 Chr.17: 18185083-18185083 on Build GRCh38
rs923372599 Chr.17: 18185141-18185141 on Build GRCh38
rs186990322 Chr.17: 18185051-18185051 on Build GRCh38
rs898861458 Chr.17: 18185072-18185072 on Build GRCh38
rs751759535 Chr.17: 18185066-18185067 on Build GRCh38
rs907644430 Chr.17: 18185032-18185032 on Build GRCh38
rs751395755 Chr.17: 18185135-18185135 on Build GRCh38
rs765189459 Chr.17: 18184981-18184981 on Build GRCh38
rs1022291692 Chr.17: 18185150-18185150 on Build GRCh38
rs985052439 Chr.17: 18185126-18185126 on Build GRCh38
rs558960178 Chr.17: 18185108-18185108 on Build GRCh38
rs971170332 Chr.17: 18185100-18185100 on Build GRCh38
rs73980817 Chr.17: 18185063-18185063 on Build GRCh38
rs1012498248 Chr.17: 18185080-18185080 on Build GRCh38
rs150152593 Chr.17: 18185103-18185103 on Build GRCh38
rs964391686 Chr.17: 18185167-18185167 on Build GRCh38
rs775566827 Chr.17: 18185055-18185055 on Build GRCh38
rs958909126 Chr.17: 18185112-18185112 on Build GRCh38
rs906680822 Chr.17: 18185091-18185091 on Build GRCh38
rs1006771506 Chr.17: 18185017-18185017 on Build GRCh38
rs751424271 Chr.17: 18185025-18185025 on Build GRCh38
rs376303822 Chr.17: 18185059-18185059 on Build GRCh38
rs117195862 Chr.17: 18185107-18185107 on Build GRCh38
rs776745020 Chr.17: 18185045-18185046 on Build GRCh38
rs759473818 Chr.17: 18185048-18185048 on Build GRCh38
rs972671010 Chr.17: 18185137-18185137 on Build GRCh38
rs541343970 Chr.17: 18185177-18185177 on Build GRCh38
rs754736495 Chr.17: 18185050-18185050 on Build GRCh38
rs766292029 Chr.17: 18185018-18185018 on Build GRCh38
rs1050345295 Chr.17: 18185173-18185173 on Build GRCh38
rs933445276 Chr.17: 18185151-18185151 on Build GRCh38
rs1003685169 Chr.17: 18185093-18185093 on Build GRCh38
rs758212755 Chr.17: 18185008-18185008 on Build GRCh38

View Full Product Details