Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73246105-73246330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC9
Entrez Gene ID
23234
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Gene Aliases
HDJC9, JDD1, SB73
UniGene
Hs.408577
Gene Chromosome Location
Chr.10: 73241954-73247331 on Build GRCh38
Transcripts
NM_015190.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6210640 c.335A>T p.D112V Chr.10: 73246163-73246163 on Build GRCh38
2161846 c.340C>G p.Q114E Chr.10: 73246158-73246158 on Build GRCh38
3415234 c.354G>T p.K118N Chr.10: 73246144-73246144 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs137976103 Chr.10: 73246161-73246161 on Build GRCh38
rs745620118 Chr.10: 73246208-73246208 on Build GRCh38
rs985000799 Chr.10: 73246281-73246281 on Build GRCh38
rs765207516 Chr.10: 73246138-73246138 on Build GRCh38
rs763077555 Chr.10: 73246163-73246163 on Build GRCh38
rs756844173 Chr.10: 73246196-73246196 on Build GRCh38
rs751535349 Chr.10: 73246169-73246170 on Build GRCh38
rs760154062 Chr.10: 73246173-73246173 on Build GRCh38
rs771929188 Chr.10: 73246130-73246130 on Build GRCh38
rs200630658 Chr.10: 73246164-73246164 on Build GRCh38
rs776722087 Chr.10: 73246134-73246134 on Build GRCh38
rs36012375 Chr.10: 73246143-73246143 on Build GRCh38
rs777144085 Chr.10: 73246191-73246191 on Build GRCh38
rs778683425 Chr.10: 73246207-73246207 on Build GRCh38
rs910841414 Chr.10: 73246283-73246283 on Build GRCh38
rs971805543 Chr.10: 73246181-73246181 on Build GRCh38
rs375346546 Chr.10: 73246188-73246188 on Build GRCh38
rs746887349 Chr.10: 73246217-73246217 on Build GRCh38
rs574062432 Chr.10: 73246271-73246271 on Build GRCh38
rs780085931 Chr.10: 73246214-73246214 on Build GRCh38
rs769978603 Chr.10: 73246220-73246220 on Build GRCh38
rs952242137 Chr.10: 73246236-73246236 on Build GRCh38
rs751910456 Chr.10: 73246165-73246165 on Build GRCh38
rs201465338 Chr.10: 73246213-73246213 on Build GRCh38
rs959173003 Chr.10: 73246230-73246230 on Build GRCh38
rs768092220 Chr.10: 73246176-73246176 on Build GRCh38
rs199733532 Chr.10: 73246136-73246136 on Build GRCh38
rs1016603322 Chr.10: 73246263-73246263 on Build GRCh38
rs990613938 Chr.10: 73246242-73246242 on Build GRCh38

View Full Product Details