Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 98307450-98307723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2L1
Entrez Gene ID
55971
Gene Name
BAI1 associated protein 2 like 1
Gene Aliases
IRTKS
UniGene
Hs.656063
Gene Chromosome Location
Chr.7: 98291650-98401115 on Build GRCh38
Transcripts
NM_018842.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

Gene Symbol
BRI3
Entrez Gene ID
25798
Gene Name
brain protein I3
Gene Aliases
I3
UniGene
Hs.567438
Gene Chromosome Location
Chr.7: 98281667-98322398 on Build GRCh38
Transcripts
NM_018842.4 XM_017011931.1 XM_017011932.1 XM_017011933.1 XM_017011934.1 XM_017011935.1 XM_017011936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4409144 c.1149C>T p.H383H Chr.7: 98307703-98307703 on Build GRCh38
5472479 c.1131C>T p.G377G Chr.7: 98307721-98307721 on Build GRCh38
4168546 c.1153G>A p.V385M Chr.7: 98307699-98307699 on Build GRCh38
7325645 c.1147delC p.H383fs*26 Chr.7: 98307705-98307705 on Build GRCh38
272382 c.1134G>T p.W378C Chr.7: 98307718-98307718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040916047 Chr.7: 98307567-98307567 on Build GRCh38
rs554719147 Chr.7: 98307539-98307539 on Build GRCh38
rs79243199 Chr.7: 98307518-98307518 on Build GRCh38
rs954345496 Chr.7: 98307603-98307603 on Build GRCh38
rs928434063 Chr.7: 98307659-98307659 on Build GRCh38
rs770765810 Chr.7: 98307701-98307701 on Build GRCh38
rs201596651 Chr.7: 98307505-98307505 on Build GRCh38
rs147254651 Chr.7: 98307700-98307700 on Build GRCh38
rs939903551 Chr.7: 98307520-98307520 on Build GRCh38
rs1057420263 Chr.7: 98307507-98307507 on Build GRCh38
rs115134774 Chr.7: 98307646-98307646 on Build GRCh38
rs371521644 Chr.7: 98307702-98307702 on Build GRCh38
rs747446690 Chr.7: 98307689-98307689 on Build GRCh38
rs762707026 Chr.7: 98307651-98307651 on Build GRCh38
rs2906187 Chr.7: 98307586-98307586 on Build GRCh38
rs199631304 Chr.7: 98307699-98307699 on Build GRCh38
rs553212732 Chr.7: 98307678-98307678 on Build GRCh38
rs11976515 Chr.7: 98307610-98307610 on Build GRCh38
rs76352870 Chr.7: 98307692-98307692 on Build GRCh38
rs368040146 Chr.7: 98307676-98307676 on Build GRCh38
rs374666080 Chr.7: 98307665-98307665 on Build GRCh38
rs780121332 Chr.7: 98307681-98307681 on Build GRCh38
rs1036717723 Chr.7: 98307527-98307527 on Build GRCh38
rs773854322 Chr.7: 98307703-98307703 on Build GRCh38
rs370945356 Chr.7: 98307664-98307664 on Build GRCh38
rs1011655233 Chr.7: 98307624-98307624 on Build GRCh38
rs766062967 Chr.7: 98307655-98307655 on Build GRCh38
rs111950058 Chr.7: 98307545-98307545 on Build GRCh38
rs1049504093 Chr.7: 98307544-98307544 on Build GRCh38
rs965583499 Chr.7: 98307652-98307652 on Build GRCh38
rs1031682761 Chr.7: 98307602-98307602 on Build GRCh38
rs781577715 Chr.7: 98307690-98307690 on Build GRCh38
rs937309659 Chr.7: 98307481-98307481 on Build GRCh38
rs541005179 Chr.7: 98307666-98307666 on Build GRCh38
rs902112650 Chr.7: 98307574-98307574 on Build GRCh38
rs1014516782 Chr.7: 98307538-98307538 on Build GRCh38
rs537246653 Chr.7: 98307554-98307554 on Build GRCh38
rs750448116 Chr.7: 98307677-98307677 on Build GRCh38
rs560898715 Chr.7: 98307471-98307471 on Build GRCh38
rs748005285 Chr.7: 98307628-98307628 on Build GRCh38
rs973028073 Chr.7: 98307686-98307686 on Build GRCh38
rs180775627 Chr.7: 98307528-98307528 on Build GRCh38

View Full Product Details