Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 10471894-10472121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APCDD1
Entrez Gene ID
147495
Gene Name
APC down-regulated 1
Gene Aliases
B7323, DRAPC1, FP7019, HHS, HTS, HYPT1
UniGene
Hs.293274
Gene Chromosome Location
Chr.18: 10454628-10488701 on Build GRCh38
Transcripts
NM_153000.4 XM_011525603.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7340297 c.719C>T p.T240I Chr.18: 10472006-10472006 on Build GRCh38
6148280 c.662A>G p.H221R Chr.18: 10471949-10471949 on Build GRCh38
159120 c.685G>C p.E229Q Chr.18: 10471972-10471972 on Build GRCh38
3989471 c.630G>A p.Q210Q Chr.18: 10471917-10471917 on Build GRCh38
437897 c.638G>A p.R213Q Chr.18: 10471925-10471925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057289531 Chr.18: 10472026-10472026 on Build GRCh38
rs117942846 Chr.18: 10472077-10472077 on Build GRCh38
rs776087296 Chr.18: 10472067-10472067 on Build GRCh38
rs752102915 Chr.18: 10472023-10472023 on Build GRCh38
rs754806469 Chr.18: 10472006-10472006 on Build GRCh38
rs778943965 Chr.18: 10472013-10472013 on Build GRCh38
rs770761989 Chr.18: 10472037-10472037 on Build GRCh38
rs936793002 Chr.18: 10472072-10472072 on Build GRCh38
rs748706455 Chr.18: 10472024-10472024 on Build GRCh38
rs1007379968 Chr.18: 10471985-10471985 on Build GRCh38
rs549800578 Chr.18: 10472041-10472041 on Build GRCh38
rs763049478 Chr.18: 10471957-10471957 on Build GRCh38
rs770344284 Chr.18: 10471948-10471948 on Build GRCh38
rs771812570 Chr.18: 10471920-10471920 on Build GRCh38
rs751758151 Chr.18: 10471988-10471988 on Build GRCh38
rs1036419184 Chr.18: 10472005-10472005 on Build GRCh38
rs764251642 Chr.18: 10471963-10471963 on Build GRCh38
rs770157936 Chr.18: 10472063-10472063 on Build GRCh38
rs763499384 Chr.18: 10472076-10472076 on Build GRCh38
rs889016458 Chr.18: 10471959-10471959 on Build GRCh38
rs771387300 Chr.18: 10471927-10471927 on Build GRCh38
rs201504676 Chr.18: 10471945-10471945 on Build GRCh38
rs537329264 Chr.18: 10471968-10471968 on Build GRCh38
rs777594465 Chr.18: 10471924-10471924 on Build GRCh38
rs777116492 Chr.18: 10471935-10471935 on Build GRCh38
rs762091049 Chr.18: 10471996-10471996 on Build GRCh38
rs780068149 Chr.18: 10472082-10472082 on Build GRCh38
rs755754147 Chr.18: 10471981-10471982 on Build GRCh38
rs897890762 Chr.18: 10472021-10472021 on Build GRCh38
rs372282253 Chr.18: 10472035-10472035 on Build GRCh38
rs571030647 Chr.18: 10472085-10472085 on Build GRCh38
rs775364287 Chr.18: 10471956-10471956 on Build GRCh38
rs746711951 Chr.18: 10471925-10471925 on Build GRCh38
rs774471766 Chr.18: 10472079-10472079 on Build GRCh38
rs746650863 Chr.18: 10472028-10472028 on Build GRCh38
rs1030269928 Chr.18: 10471953-10471953 on Build GRCh38
rs139232963 Chr.18: 10472001-10472001 on Build GRCh38
rs368209332 Chr.18: 10471999-10471999 on Build GRCh38
rs980981047 Chr.18: 10472061-10472061 on Build GRCh38
rs777408847 Chr.18: 10472013-10472015 on Build GRCh38

View Full Product Details