Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232456281-232456540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALPI
Entrez Gene ID
248
Gene Name
alkaline phosphatase, intestinal
Gene Aliases
IAP
UniGene
Hs.37009
Gene Chromosome Location
Chr.2: 232456112-232460745 on Build GRCh38
Transcripts
NM_001631.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598741 c.97C>T p.R33C Chr.2: 232456378-232456378 on Build GRCh38
3579081 c.113C>T p.A38V Chr.2: 232456394-232456394 on Build GRCh38
7129342 c.100C>G p.Q34E Chr.2: 232456381-232456381 on Build GRCh38
4661220 c.106G>A p.A36T Chr.2: 232456387-232456387 on Build GRCh38
3798747 c.102G>A p.Q34Q Chr.2: 232456383-232456383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766664750 Chr.2: 232456494-232456494 on Build GRCh38
rs553121411 Chr.2: 232456425-232456425 on Build GRCh38
rs750797497 Chr.2: 232456388-232456388 on Build GRCh38
rs764209297 Chr.2: 232456378-232456378 on Build GRCh38
rs558991529 Chr.2: 232456516-232456516 on Build GRCh38
rs376733154 Chr.2: 232456335-232456335 on Build GRCh38
rs201573533 Chr.2: 232456364-232456364 on Build GRCh38
rs752096410 Chr.2: 232456314-232456314 on Build GRCh38
rs768049873 Chr.2: 232456383-232456383 on Build GRCh38
rs757651183 Chr.2: 232456318-232456318 on Build GRCh38
rs929275351 Chr.2: 232456374-232456374 on Build GRCh38
rs937914640 Chr.2: 232456338-232456338 on Build GRCh38
rs748141097 Chr.2: 232456419-232456419 on Build GRCh38
rs1055016246 Chr.2: 232456342-232456342 on Build GRCh38
rs745561957 Chr.2: 232456333-232456333 on Build GRCh38
rs779654521 Chr.2: 232456339-232456339 on Build GRCh38
rs756498447 Chr.2: 232456390-232456390 on Build GRCh38
rs752666286 Chr.2: 232456405-232456418 on Build GRCh38
rs1038189727 Chr.2: 232456344-232456344 on Build GRCh38
rs376557863 Chr.2: 232456509-232456509 on Build GRCh38
rs761925584 Chr.2: 232456466-232456466 on Build GRCh38
rs778354363 Chr.2: 232456414-232456414 on Build GRCh38
rs771952254 Chr.2: 232456424-232456424 on Build GRCh38
rs759327136 Chr.2: 232456351-232456353 on Build GRCh38
rs778604684 Chr.2: 232456514-232456514 on Build GRCh38
rs768884728 Chr.2: 232456461-232456461 on Build GRCh38
rs369242928 Chr.2: 232456470-232456470 on Build GRCh38
rs774242869 Chr.2: 232456462-232456462 on Build GRCh38
rs948606722 Chr.2: 232456325-232456325 on Build GRCh38
rs772832583 Chr.2: 232456358-232456358 on Build GRCh38
rs774077251 Chr.2: 232456335-232456336 on Build GRCh38
rs771934296 Chr.2: 232456345-232456345 on Build GRCh38
rs551044260 Chr.2: 232456301-232456301 on Build GRCh38
rs201034820 Chr.2: 232456362-232456362 on Build GRCh38
rs775849282 Chr.2: 232456365-232456365 on Build GRCh38
rs775865612 Chr.2: 232456435-232456435 on Build GRCh38
rs566739473 Chr.2: 232456443-232456443 on Build GRCh38
rs1011106503 Chr.2: 232456482-232456482 on Build GRCh38
rs752873124 Chr.2: 232456306-232456306 on Build GRCh38
rs957559574 Chr.2: 232456487-232456487 on Build GRCh38
rs959965735 Chr.2: 232456304-232456304 on Build GRCh38
rs752188879 Chr.2: 232456515-232456515 on Build GRCh38
rs758519310 Chr.2: 232456308-232456308 on Build GRCh38
rs748867233 Chr.2: 232456340-232456340 on Build GRCh38
rs748883641 Chr.2: 232456399-232456399 on Build GRCh38
rs776916372 Chr.2: 232456300-232456300 on Build GRCh38
rs777628025 Chr.2: 232456517-232456517 on Build GRCh38
rs774286323 Chr.2: 232456341-232456341 on Build GRCh38
rs777726761 Chr.2: 232456510-232456510 on Build GRCh38
rs144782038 Chr.2: 232456492-232456492 on Build GRCh38
rs202137698 Chr.2: 232456434-232456434 on Build GRCh38
rs1033193145 Chr.2: 232456475-232456475 on Build GRCh38
rs761044216 Chr.2: 232456493-232456493 on Build GRCh38
rs781341355 Chr.2: 232456332-232456332 on Build GRCh38
rs61732026 Chr.2: 232456379-232456379 on Build GRCh38
rs763180891 Chr.2: 232456369-232456369 on Build GRCh38
rs764763129 Chr.2: 232456312-232456312 on Build GRCh38
rs900665350 Chr.2: 232456472-232456472 on Build GRCh38
rs767398322 Chr.2: 232456355-232456355 on Build GRCh38
rs779742623 Chr.2: 232456394-232456394 on Build GRCh38

View Full Product Details