Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 62603314-62603587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01465
Entrez Gene ID
283416
Gene Name
long intergenic non-protein coding RNA 1465
Gene Aliases
C12orf61
UniGene
Hs.375881
Gene Chromosome Location
Chr.12: 62601751-62603434 on Build GRCh38
Transcripts
NR_121682.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6381518 c.7G>T p.G3C Chr.12: 62603332-62603332 on Build GRCh38
4627130 c.16G>A p.A6T Chr.12: 62603323-62603323 on Build GRCh38
4774169 c.19G>T p.V7F Chr.12: 62603320-62603320 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1033929277 Chr.12: 62603374-62603374 on Build GRCh38
rs547302253 Chr.12: 62603397-62603397 on Build GRCh38
rs370685868 Chr.12: 62603417-62603417 on Build GRCh38
rs944627822 Chr.12: 62603426-62603426 on Build GRCh38
rs953908445 Chr.12: 62603479-62603479 on Build GRCh38
rs1056650351 Chr.12: 62603415-62603415 on Build GRCh38
rs536001911 Chr.12: 62603402-62603402 on Build GRCh38
rs961327234 Chr.12: 62603471-62603471 on Build GRCh38
rs749562233 Chr.12: 62603411-62603411 on Build GRCh38
rs752377300 Chr.12: 62603333-62603333 on Build GRCh38
rs572685822 Chr.12: 62603525-62603525 on Build GRCh38
rs559310321 Chr.12: 62603497-62603497 on Build GRCh38
rs887756665 Chr.12: 62603444-62603444 on Build GRCh38
rs924147793 Chr.12: 62603564-62603564 on Build GRCh38
rs1040155553 Chr.12: 62603432-62603432 on Build GRCh38
rs796184182 Chr.12: 62603400-62603401 on Build GRCh38
rs922323901 Chr.12: 62603348-62603348 on Build GRCh38
rs186694256 Chr.12: 62603566-62603566 on Build GRCh38
rs985157525 Chr.12: 62603483-62603483 on Build GRCh38
rs895210928 Chr.12: 62603422-62603422 on Build GRCh38
rs939464186 Chr.12: 62603401-62603401 on Build GRCh38
rs985903171 Chr.12: 62603414-62603414 on Build GRCh38
rs766726101 Chr.12: 62603380-62603380 on Build GRCh38
rs10877887 Chr.12: 62603400-62603400 on Build GRCh38
rs556257065 Chr.12: 62603442-62603442 on Build GRCh38
rs946426558 Chr.12: 62603393-62603393 on Build GRCh38
rs914877477 Chr.12: 62603384-62603384 on Build GRCh38
rs954271945 Chr.12: 62603532-62603532 on Build GRCh38
rs945236906 Chr.12: 62603437-62603437 on Build GRCh38
rs538612367 Chr.12: 62603474-62603474 on Build GRCh38
rs1029462987 Chr.12: 62603477-62603477 on Build GRCh38
rs897313369 Chr.12: 62603495-62603495 on Build GRCh38
rs757204594 Chr.12: 62603366-62603366 on Build GRCh38
rs113188524 Chr.12: 62603373-62603373 on Build GRCh38
rs576113439 Chr.12: 62603450-62603450 on Build GRCh38
rs1012353015 Chr.12: 62603441-62603441 on Build GRCh38
rs1027067357 Chr.12: 62603518-62603518 on Build GRCh38
rs1015918983 Chr.12: 62603457-62603457 on Build GRCh38
rs961290113 Chr.12: 62603376-62603376 on Build GRCh38
rs1004925218 Chr.12: 62603452-62603452 on Build GRCh38
rs867804200 Chr.12: 62603511-62603511 on Build GRCh38
rs763658721 Chr.12: 62603345-62603345 on Build GRCh38
rs113451066 Chr.12: 62603418-62603418 on Build GRCh38
rs954063919 Chr.12: 62603405-62603405 on Build GRCh38
rs182504454 Chr.12: 62603557-62603557 on Build GRCh38
rs1055204401 Chr.12: 62603454-62603454 on Build GRCh38
rs534077094 Chr.12: 62603385-62603385 on Build GRCh38
rs752740994 Chr.12: 62603424-62603424 on Build GRCh38
rs1043350801 Chr.12: 62603346-62603346 on Build GRCh38
rs760425891 Chr.12: 62603336-62603336 on Build GRCh38
rs968108123 Chr.12: 62603544-62603544 on Build GRCh38
rs541690178 Chr.12: 62603541-62603541 on Build GRCh38
rs913755984 Chr.12: 62603435-62603435 on Build GRCh38
rs903636850 Chr.12: 62603446-62603446 on Build GRCh38
rs753404213 Chr.12: 62603353-62603353 on Build GRCh38

View Full Product Details