Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53424672-53424929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMHR2
Entrez Gene ID
269
Gene Name
anti-Mullerian hormone receptor type 2
Gene Aliases
AMHR, MISR2, MISRII, MRII
UniGene
Hs.659889
Gene Chromosome Location
Chr.12: 53423855-53431672 on Build GRCh38
Transcripts
NM_001164690.1 NM_001164691.1 NM_020547.2 XM_011538173.1 XM_011538174.1 XM_011538176.1 XM_011538178.1 XM_011538179.1 XM_011538180.1 XM_011538181.1 XM_011538183.2 XM_011538184.2 XM_011538185.1 XM_011538187.2 XM_017019179.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6299243 c.238C>T p.R80* Chr.12: 53424714-53424714 on Build GRCh38
4575551 c.338C>T p.T113I Chr.12: 53424814-53424814 on Build GRCh38
3462673 c.374C>T p.P125L Chr.12: 53424850-53424850 on Build GRCh38
1933034 c.424G>T p.G142C Chr.12: 53424900-53424900 on Build GRCh38
1933032 c.322A>C p.T108P Chr.12: 53424798-53424798 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147584433 Chr.12: 53424803-53424803 on Build GRCh38
rs768584073 Chr.12: 53424744-53424744 on Build GRCh38
rs768162879 Chr.12: 53424821-53424821 on Build GRCh38
rs772348472 Chr.12: 53424887-53424887 on Build GRCh38
rs750370851 Chr.12: 53424877-53424878 on Build GRCh38
rs778758485 Chr.12: 53424706-53424706 on Build GRCh38
rs777559665 Chr.12: 53424827-53424827 on Build GRCh38
rs929791810 Chr.12: 53424886-53424886 on Build GRCh38
rs373234304 Chr.12: 53424765-53424765 on Build GRCh38
rs768744210 Chr.12: 53424843-53424843 on Build GRCh38
rs758474279 Chr.12: 53424882-53424883 on Build GRCh38
rs753454794 Chr.12: 53424697-53424697 on Build GRCh38
rs911169796 Chr.12: 53424826-53424826 on Build GRCh38
rs369659478 Chr.12: 53424739-53424739 on Build GRCh38
rs376565347 Chr.12: 53424714-53424714 on Build GRCh38
rs564662907 Chr.12: 53424762-53424762 on Build GRCh38
rs760383339 Chr.12: 53424769-53424769 on Build GRCh38
rs896993874 Chr.12: 53424885-53424885 on Build GRCh38
rs774355890 Chr.12: 53424845-53424845 on Build GRCh38
rs774586083 Chr.12: 53424749-53424749 on Build GRCh38
rs187185951 Chr.12: 53424908-53424908 on Build GRCh38
rs1040945449 Chr.12: 53424881-53424881 on Build GRCh38
rs779678943 Chr.12: 53424884-53424886 on Build GRCh38
rs967541474 Chr.12: 53424728-53424728 on Build GRCh38
rs762025424 Chr.12: 53424867-53424867 on Build GRCh38
rs758069288 Chr.12: 53424805-53424805 on Build GRCh38
rs771456549 Chr.12: 53424713-53424713 on Build GRCh38
rs764851153 Chr.12: 53424816-53424818 on Build GRCh38
rs770395435 Chr.12: 53424726-53424726 on Build GRCh38
rs775842856 Chr.12: 53424733-53424733 on Build GRCh38
rs957579736 Chr.12: 53424719-53424719 on Build GRCh38
rs377707786 Chr.12: 53424892-53424892 on Build GRCh38
rs770030378 Chr.12: 53424833-53424833 on Build GRCh38
rs747774459 Chr.12: 53424711-53424711 on Build GRCh38
rs141999682 Chr.12: 53424828-53424828 on Build GRCh38
rs766399797 Chr.12: 53424775-53424775 on Build GRCh38
rs761339798 Chr.12: 53424755-53424762 on Build GRCh38
rs753698788 Chr.12: 53424785-53424785 on Build GRCh38
rs767312699 Chr.12: 53424763-53424763 on Build GRCh38
rs753464279 Chr.12: 53424737-53424738 on Build GRCh38
rs552083814 Chr.12: 53424798-53424798 on Build GRCh38
rs749717207 Chr.12: 53424838-53424838 on Build GRCh38
rs202187359 Chr.12: 53424898-53424898 on Build GRCh38
rs746378444 Chr.12: 53424725-53424725 on Build GRCh38
rs754434471 Chr.12: 53424702-53424702 on Build GRCh38
rs751416712 Chr.12: 53424890-53424894 on Build GRCh38
rs761735257 Chr.12: 53424715-53424715 on Build GRCh38
rs534750443 Chr.12: 53424836-53424836 on Build GRCh38

View Full Product Details