Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 10642772-10643019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASZ1
Entrez Gene ID
54897
Gene Name
castor zinc finger 1
Gene Aliases
CAS11, CST, SRG, ZNF693, dJ734G22.1
UniGene
Hs.439894
Gene Chromosome Location
Chr.1: 10636602-10796676 on Build GRCh38
Transcripts
NM_001079843.2 XM_005263479.3 XM_017001539.1 XM_017001540.1 XM_017001541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4020086 c.4025C>A p.P1342H Chr.1: 10642996-10642996 on Build GRCh38
6057156 c.4138G>T p.V1380L Chr.1: 10642883-10642883 on Build GRCh38
6364897 c.4130G>T p.S1377I Chr.1: 10642891-10642891 on Build GRCh38
3470583 c.4118G>A p.R1373Q Chr.1: 10642903-10642903 on Build GRCh38
2116243 c.4068C>G p.Y1356* Chr.1: 10642953-10642953 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953224837 Chr.1: 10642985-10642985 on Build GRCh38
rs766224188 Chr.1: 10642970-10642970 on Build GRCh38
rs773910498 Chr.1: 10642945-10642945 on Build GRCh38
rs773373628 Chr.1: 10642816-10642816 on Build GRCh38
rs539367148 Chr.1: 10642836-10642836 on Build GRCh38
rs576145769 Chr.1: 10642959-10642959 on Build GRCh38
rs746292412 Chr.1: 10642915-10642915 on Build GRCh38
rs369880059 Chr.1: 10642852-10642852 on Build GRCh38
rs750391884 Chr.1: 10642961-10642961 on Build GRCh38
rs770207190 Chr.1: 10642934-10642934 on Build GRCh38
rs758520250 Chr.1: 10642823-10642824 on Build GRCh38
rs778124789 Chr.1: 10643000-10643000 on Build GRCh38
rs755943898 Chr.1: 10642874-10642874 on Build GRCh38
rs903417536 Chr.1: 10642927-10642927 on Build GRCh38
rs894686919 Chr.1: 10642869-10642869 on Build GRCh38
rs375809156 Chr.1: 10642883-10642883 on Build GRCh38
rs200729371 Chr.1: 10642849-10642849 on Build GRCh38
rs1025967736 Chr.1: 10642894-10642894 on Build GRCh38
rs537672299 Chr.1: 10642969-10642969 on Build GRCh38
rs373145502 Chr.1: 10642865-10642865 on Build GRCh38
rs979864141 Chr.1: 10642960-10642960 on Build GRCh38
rs777190275 Chr.1: 10642910-10642910 on Build GRCh38
rs772294858 Chr.1: 10642810-10642810 on Build GRCh38
rs750255554 Chr.1: 10642872-10642872 on Build GRCh38
rs758740834 Chr.1: 10642992-10642992 on Build GRCh38
rs763715781 Chr.1: 10642939-10642939 on Build GRCh38
rs550410948 Chr.1: 10642801-10642801 on Build GRCh38
rs115628512 Chr.1: 10642884-10642884 on Build GRCh38
rs367856340 Chr.1: 10642845-10642845 on Build GRCh38
rs753650518 Chr.1: 10642976-10642976 on Build GRCh38
rs747059584 Chr.1: 10642903-10642903 on Build GRCh38
rs368787986 Chr.1: 10642863-10642863 on Build GRCh38
rs199766638 Chr.1: 10642870-10642870 on Build GRCh38
rs376223327 Chr.1: 10642999-10642999 on Build GRCh38
rs763627573 Chr.1: 10642853-10642853 on Build GRCh38
rs757234674 Chr.1: 10642864-10642864 on Build GRCh38
rs765197520 Chr.1: 10642982-10642982 on Build GRCh38
rs778034955 Chr.1: 10642899-10642899 on Build GRCh38
rs761311758 Chr.1: 10642958-10642958 on Build GRCh38
rs376551786 Chr.1: 10642821-10642821 on Build GRCh38
rs183558878 Chr.1: 10642935-10642935 on Build GRCh38

View Full Product Details